Skip to Content
Merck
All Photos(1)

Documents

343536

Sigma-Aldrich

1-Boc-piperazine

97%

Synonym(s):

tert-Butyl piperazine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2O2
CAS Number:
Molecular Weight:
186.25
Beilstein:
879985
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

43-47 °C (lit.)

SMILES string

CC(C)(C)OC(=O)N1CCNCC1

InChI

1S/C9H18N2O2/c1-9(2,3)13-8(12)11-6-4-10-5-7-11/h10H,4-7H2,1-3H3

InChI key

CWXPZXBSDSIRCS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Boc-piperazine is an N-Boc protected piperazine. Crosscoupling of 1-Boc-piperazine with aryl iodides using CuBr/1,1′-bi-2-naphthol (catalyst) and K3PO4 (base) has been reported. 1-Boc-piperazine undergoes Buchwald-Hartwig coupling reactions with aryl halides. 1-Boc-piperazine can be prepared in 80% yield via solvent-free N-Boc protection catalyzed by iodine.

Application

1-Boc-piperazine may be used in:
  • preparation of series of (m-phenoxy)phenyl substituted piperazine derivatives
  • termination step during synthesis of α,β-poly(2-oxazoline) lipopolymers via living cationic ring opening polymerization
  • preparation of monosubstituted piperazines, e.g. in the synthesis of indazole DNA gyrase inhibitors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dengfeng Dou et al.
Bioorganic & medicinal chemistry letters, 22(1), 377-379 (2011-11-29)
There is currently an unmet need for the development of small-molecule therapeutics for norovirus infection. The piperazine scaffold, a privileged structure embodied in many pharmacological agents, was used to synthesize an array of structurally-diverse derivatives which were screened for anti-norovius
Akihiko Tanitame et al.
Bioorganic & medicinal chemistry letters, 14(11), 2857-2862 (2004-05-06)
In this study, we report the design, synthesis and structure-activity relationships of novel indazole derivatives as DNA gyrase inhibitors with Gram-positive antibacterial activity. Our results show that selected compounds from this series exhibit potent antibacterial activity against Gram-positive bacteria including
Florencio Zaragoza et al.
Journal of medicinal chemistry, 47(11), 2833-2838 (2004-05-14)
With the aim of identifying structurally novel, centrally acting histamine H(3) antagonists, arrays of monoacyldiamines were screened. This led to the discovery of a series of 1-alkyl-4-acylpiperazines which were potent antagonists at the human histamine H(3) receptor. The most potent
α,ω-Functionalized Poly (2-Oxazoline) s Bearing Hydroxyl and Amino Functions.
Reif M and Jordan R.
Macromolecular Chemistry and Physics, 212(16), 1815-1824 (2011)
Ravi Varala et al.
The Journal of organic chemistry, 71(21), 8283-8286 (2006-10-10)
An efficient and practical protocol for the protection of various structurally and electronically divergent aryl and aliphatic amines using (Boc)2O in the presence of a catalytic amount of molecular iodine (10 mol %) under solvent-free conditions at ambient temperature is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service