695599
2-(2-(Diphenylphosphino)ethyl)pyridine
kanata purity
Synonym(s):
1-(Diphenylphosphino)-2-(2-pyridyl)ethane, Diphenyl(2-(pyridin-2-yl)ethyl)phosphine
About This Item
Recommended Products
form
solid
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
mp
58-62 °C
functional group
phosphine
SMILES string
C(Cc1ccccn1)P(c2ccccc2)c3ccccc3
InChI
1S/C19H18NP/c1-3-10-18(11-4-1)21(19-12-5-2-6-13-19)16-14-17-9-7-8-15-20-17/h1-13,15H,14,16H2
InChI key
ARSGXAZDYSTSKI-UHFFFAOYSA-N
Related Categories
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service