Skip to Content
Merck
All Photos(2)

Documents

392960

Sigma-Aldrich

Diphenyl-2-pyridylphosphine

97%

Synonym(s):

2-(Diphenylphosphino)pyridine, 2-Pyridyldiphenylphosphine, Diphenyl(2-pyridinyl)phosphine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14NP
CAS Number:
Molecular Weight:
263.27
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reagent type: ligand
reaction type: Carbonatations

reagent type: ligand
reaction type: Dehydrogenation

reagent type: ligand
reaction type: Hydration Reaction

reagent type: ligand
reaction type: Mitsunobu Reaction

reagent type: ligand
reaction type: Stannylation

impurities

1-3% 2-bromopyridine

mp

82-84 °C (dec.) (lit.)

functional group

phosphine

SMILES string

c1ccc(cc1)P(c2ccccc2)c3ccccn3

InChI

1S/C17H14NP/c1-3-9-15(10-4-1)19(16-11-5-2-6-12-16)17-13-7-8-14-18-17/h1-14H

InChI key

SVABQOITNJTVNJ-UHFFFAOYSA-N

Application

Ligand for metal-catalyzed carbonylations, hydration, dehydrogenative coupling, carbostannylation, distannylation, and silylation; reagent for Mitsunobu reaction.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroto Yoshida et al.
Organic letters, 8(18), 4157-4159 (2006-08-25)
An exo-diene moiety of various ortho-quinodimethanes, regardless of its transient character, was inserted into a Sn-Sn sigma-bond of hexabutyldistannane in the presence of a palladium catalyst, giving alpha,alpha'-bis(tributylstannyl)-o-xylenes straightforwardly.
Oshiki, T.; Yamashita, H. et al.
Organometallics, 24, 6287-6287 (2005)
Nickel-catalyzed tandem carbostannylation of alkynes and 1,2-dienes with alkynylstannanes.
Eiji Shirakawa et al.
Angewandte Chemie (International ed. in English), 43(26), 3448-3451 (2004-06-29)
Consorti, C. S.; Ebeling, G.; Dupont, J.
Tetrahedron Letters, 43, 753-753 (2002)
Xianjin Lan et al.
Journal of hazardous materials, 375, 26-32 (2019-04-30)
The worldwide environmental occurrence of natural steroid estrogens has drawn increasing concerns. However, the fate of the estrogens, especially the α-isomer of estradiol, in the environmental matrices is still obscure. Using 14C-radioactively labelled forms of these estrogens can facilitate and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service