Skip to Content
Merck
All Photos(1)

Key Documents

682136

Sigma-Aldrich

(S)-(+)-4,12-Bis(diphenylphosphino)-[2.2]-paracyclophane

96%

Synonym(s):

(S)-Phanephos

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H34P2
CAS Number:
Molecular Weight:
576.65
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

form

solid

optical activity

[α]22/D +34°, c = 1 in chloroform

mp

224-226 °C

SMILES string

C1Cc2ccc(CCc3ccc1cc3P(c4ccccc4)c5ccccc5)cc2P(c6ccccc6)c7ccccc7

InChI

1S/C40H34P2/c1-5-13-35(14-6-1)41(36-15-7-2-8-16-36)39-29-31-21-25-33(39)27-23-32-22-26-34(28-24-31)40(30-32)42(37-17-9-3-10-18-37)38-19-11-4-12-20-38/h1-22,25-26,29-30H,23-24,27-28H2

InChI key

GYZZZILPVUYAFJ-UHFFFAOYSA-N

Application

Efficient ligand for asymmetric hydrogentation of dehydroamino acids, methyl esters and keytones.

Legal Information

Sold in collaboration with Johnson Matthey Catalyst for research purposes only. US5874629 and any patents arising therefrom apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 119, 6207-6207 (1997)
Burk et al.
Organic letters, 2(26), 4173-4176 (2001-01-11)
PhanePhos-ruthenium-diamine complexes catalyze the asymmetric hydrogenation of a wide range of aromatic, heteroaromatic, and alpha, beta-unsaturated ketones with high activity and excellent enantioselectivity.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service