667854
(+)-1,2-Bis((2S,5S)-2,5-diphenylphospholano)ethane
kanata purity
Synonym(s):
(S,S)-Ph-BPE,BPE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C34H36P2
CAS Number:
Molecular Weight:
506.60
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
form
solid
mp
143-147 °C
functional group
phenyl
phosphine
SMILES string
C1C[C@H](P(CCP2[C@@H](CC[C@H]2c3ccccc3)c4ccccc4)[C@@H]1c5ccccc5)c6ccccc6
InChI
1S/C34H36P2/c1-5-13-27(14-6-1)31-21-22-32(28-15-7-2-8-16-28)35(31)25-26-36-33(29-17-9-3-10-18-29)23-24-34(36)30-19-11-4-12-20-30/h1-20,31-34H,21-26H2/t31-,32-,33-,34-/m0/s1
InChI key
VHHAZLMVLLIMHT-CUPIEXAXSA-N
Related Categories
General description
Chiral Phosphine ligand used in the asymmetric hydrogenation of allylic sulfoxides
Dynamic Kinetic Resolution of Allylic Sulfoxides by Rh-Catalyzed Hydrogenation: A Combined Theoretical and Experimental Mechanistic Study
Dynamic Kinetic Resolution of Allylic Sulfoxides by Rh-Catalyzed Hydrogenation: A Combined Theoretical and Experimental Mechanistic Study
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service