Skip to Content
Merck
All Photos(1)

Key Documents

674656

Sigma-Aldrich

(R)-(+)-2-Methyl-CBS-oxazaborolidine solution

1 M in THF

Synonym(s):

α,α-Diphenyl-D-prolinolmethylboronic acid cyclamide ester, (R)-1-Methyl,3,3-diphenyl-tetrahydro-pyrrolo(1,2-c)(1,3,2)oxazaborole, (R)-Tetrahydro-1-methyl-3,3-diphenyl-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20BNO
CAS Number:
Molecular Weight:
277.17
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

optical activity

[α]22/D +76.8°, c = 1 in toluene

concentration

1 M in THF

refractive index

n20/D 1.4602

density

0.942 g/mL at 25 °C

functional group

phenyl

SMILES string

[H][C@]12CCCN1B(C)OC2(c3ccccc3)c4ccccc4

InChI

1S/C18H20BNO/c1-19-20-14-8-13-17(20)18(21-19,15-9-4-2-5-10-15)16-11-6-3-7-12-16/h2-7,9-12,17H,8,13-14H2,1H3/t17-/m1/s1

InChI key

VMKAFJQFKBASMU-QGZVFWFLSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The CBS (Corey-Bakshi-Shibata) oxazaborolidine catalyst has been used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2 symmetrical ferrocenyl diols, and propargyl alcohols.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stemmler, R.T.
Synlett, 997-997 (2007)
Current Science, 78, 1314-1317 (2000)
Singh, V. K.
Synthesis, 605-605 (1992)
Corey, E. J. et al.
Journal of the American Chemical Society, 109, 5551-5551 (1987)
Asymmetric Reduction. A Convenient Method for the Reduction of Alkynyl Ketones.
Kathlyn A. Parker et al.
The Journal of organic chemistry, 61(9), 3214-3217 (1996-05-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service