Skip to Content
Merck
All Photos(1)

Key Documents

649317

Sigma-Aldrich

(R)-(+)-2-Methyl-CBS-oxazaborolidine

Synonym(s):

(R)-1-Methyl-3,3-diphenylhexahydropyrrolo[1,2-c][1,3,2]oxazaborole, (R)-3,3-Diphenyl-1-methylpyrrolidino[1,2-c]-1,3,2-oxazaborole, (R)-Me-Corey-Bakshi-Shibata catalyst

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H20BNO
CAS Number:
Molecular Weight:
277.17
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥90%

form

solid

mp

85-95 °C (lit.)

SMILES string

CB1OC(C2CCCN12)(c3ccccc3)c4ccccc4

InChI

1S/C18H20BNO/c1-19-20-14-8-13-17(20)18(21-19,15-9-4-2-5-10-15)16-11-6-3-7-12-16/h2-7,9-12,17H,8,13-14H2,1H3/t17-/m1/s1

InChI key

VMKAFJQFKBASMU-QGZVFWFLSA-N

Application

The CBS (Corey-Bakshi-Shibata) oxazaborolidine catalyst has been used in the asymmetric reduction of prochiral ketones. Other applications include the enantioselective synthesis of α-hydroxy acids, α-amino acids, C2 symmetrical ferrocenyl diols, and propargyl alcohols.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sing, V. K.
Synthesis, 605-605 (1992)
Corey, E. J. et al.
Journal of the American Chemical Society, 109, 5551-5551 (1987)
Schwink, L.; Knochel, P.
Tetrahedron Letters, 37, 25-25 (1996)
Corey, E. J.; Link, J. O.
Tetrahedron Letters, 33, 3431-3431 (1992)
Asymmetric Reduction. A Convenient Method for the Reduction of Alkynyl Ketones.
Kathlyn A. Parker et al.
The Journal of organic chemistry, 61(9), 3214-3217 (1996-05-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service