663085
(5S)-(−)-2,2,3-Trimethyl-5-benzyl-4-imidazolidinone dichloroacetic acid
97%
Synonym(s):
(5S)-2,2,3-Trimethyl-5-phenylmethyl-4-imidazolidinone dichloroacetic acid
About This Item
Recommended Products
Assay
97%
mp
122-126 °C
functional group
carboxylic acid
chloro
phenyl
SMILES string
OC(=O)C(Cl)Cl.CN1C(=O)[C@H](Cc2ccccc2)NC1(C)C
InChI
1S/C13H18N2O.C2H2Cl2O2/c1-13(2)14-11(12(16)15(13)3)9-10-7-5-4-6-8-10;3-1(4)2(5)6/h4-8,11,14H,9H2,1-3H3;1H,(H,5,6)/t11-;/m0./s1
InChI key
YZRAPAXOLQHZSE-MERQFXBCSA-N
Application
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service