661902
(S)-(+)-2-(tert-Butyl)-3-methyl-4-imidazolidinone trifluoroacetic acid
96%
Synonym(s):
(S)-(+)-2-(tert-Butyl)-3-methyl-4-oxoimidazolidinium trifluoroacetate, (S)-2-(tert-Butyl)-3-methyl-4-imidazolidinone trifluoroacetic acid salt
About This Item
Recommended Products
Assay
96%
form
solid
mp
99-103 °C
functional group
carboxylic acid
fluoro
SMILES string
OC(=O)C(F)(F)F.CN1[C@H](NCC1=O)C(C)(C)C
InChI
1S/C8H16N2O.C2HF3O2/c1-8(2,3)7-9-5-6(11)10(7)4;3-2(4,5)1(6)7/h7,9H,5H2,1-4H3;(H,6,7)/t7-;/m0./s1
InChI key
HKHKOEMMPVPVOS-FJXQXJEOSA-N
General description
Application
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service