Skip to Content
Merck
All Photos(2)

Key Documents

662283

Sigma-Aldrich

Bis(tert-butylcarbonyloxy)iodobenzene

97%

Synonym(s):

(Di-tert-butylcarbonyloxyiodo)benzol, 2,2-Dimethylpropanoic acid, phenyliodine complex, Bis(2,2-dimethylpropanoato-O)phenyliodide, Di-(Pivaloyloxy)iodobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H23IO4
CAS Number:
Molecular Weight:
406.26
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

reaction suitability

reagent type: catalyst
reaction type: C-H Activation

reagent type: oxidant

mp

104-109 °C

functional group

iodo

SMILES string

CC(C)(C)C(=O)O[I](OC(=O)C(C)(C)C)c1ccccc1

InChI

1S/C16H23IO4/c1-15(2,3)13(18)20-17(12-10-8-7-9-11-12)21-14(19)16(4,5)6/h7-11H,1-6H3

InChI key

DZKPLZUSZXYHFB-UHFFFAOYSA-N

Application

Rh2(esp)2: An Exceptionally Efficient and Selective Catalyst for C-H Amination

Reagent for:
  • Preparation of α,β-unsaturated-γ-lactams via Rh-catalyzed C-H amination of allene carbamates, then Ru-catalyzed cyclocarbonylation
  • Palladium-catalyzed diamination
  • Preparation of functionalized bicyclic heterocyclic compounds by rhodium-catalyzed allene amidation and cyclization
  • Preparation of piperidine and pyrrolidine derivatives via copper-catalyzed intramolecular aminoacetoxylation of aminoolefins
  • C-H acyloxylation of arenes

  • Hypervalent iodine oxidant
Hypervalent iodine reagent used with the Rh2(esp)2 catalyst (662623) in the amination of C-H bonds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dzmitry M Zubrytski et al.
Molecules (Basel, Switzerland), 26(1) (2021-01-06)
Oxidative fragmentation of tertiary cyclopropanols with phenyliodine(III) dicarboxylates in aprotic solvents (dichloromethane, chloroform, toluene) produces mixed anhydrides. The fragmentation reaction is especially facile with phenyliodine(III) reagents bearing electron-withdrawing carboxylate ligands (trifluoroacetyl, 2,4,6-trichlorobenzoyl, 3-nitrobenzoyl), and affords 95-98% yields of the corresponding
Kristin Williams Fiori et al.
Journal of the American Chemical Society, 129(3), 562-568 (2007-01-18)
Reaction methodology for intermolecular C-H amination of benzylic and 3 degrees C-H bonds is described. This process uses the starting alkane as the limiting reagent, gives optically pure tetrasubstituted amines through stereospecific insertion into enantiomeric 3 degrees centers, displays high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service