Skip to Content
Merck
All Photos(3)

Key Documents

662623

Sigma-Aldrich

Bis[rhodium(α,α,α′,α′-tetramethyl-1,3-benzenedipropionic acid)]

95%

Synonym(s):

Rh2(esp)2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H40O8Rh2
CAS Number:
Molecular Weight:
758.47
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

reaction suitability

core: rhodium
reagent type: catalyst
reaction type: C-H Activation

mp

>300 °C

SMILES string

[Rh][Rh].CC(C)(Cc1cccc(CC(C)(C)C(O)=O)c1)C(O)=O.CC(C)(Cc2cccc(CC(C)(C)C(O)=O)c2)C(O)=O

InChI

1S/2C16H22O4.2Rh/c2*1-15(2,13(17)18)9-11-6-5-7-12(8-11)10-16(3,4)14(19)20;;/h2*5-8H,9-10H2,1-4H3,(H,17,18)(H,19,20);;

InChI key

OBMUTUNJWNQIAJ-UHFFFAOYSA-N

General description

Bis[rhodium(α,α,α′,α′-tetramethyl-1,3-benzenedipropionicacid)] has been identified as efficient C-H activation catalyst.

Application

Bis[rhodium(α,α,α′,α′-tetramethyl-1,3-benzenedipropionicacid)] is used as a catalyst for oxidative C−H amination of urea and guanidinederived substrates to yield corresponding heterocyclic products.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kristin Williams Fiori et al.
Journal of the American Chemical Society, 129(3), 562-568 (2007-01-18)
Reaction methodology for intermolecular C-H amination of benzylic and 3 degrees C-H bonds is described. This process uses the starting alkane as the limiting reagent, gives optically pure tetrasubstituted amines through stereospecific insertion into enantiomeric 3 degrees centers, displays high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service