Skip to Content
Merck
All Photos(1)

Documents

632317

Sigma-Aldrich

Indium(III) oxide

nanopowder, <100 nm particle size (TEM), 99.9% trace metals basis

Synonym(s):

Diindium trioxide, Indium sesquioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
In2O3
CAS Number:
Molecular Weight:
277.63
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

<0.01 mmHg ( 25 °C)

Quality Level

Assay

99.9% trace metals basis

form

nanopowder

reaction suitability

reagent type: catalyst
core: indium

particle size

<100 nm (TEM)

density

7.18 g/mL at 25 °C (lit.)

SMILES string

O=[In]O[In]=O

InChI

1S/2In.3O

InChI key

SHTGRZNPWBITMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Indium(III)oxide is a versatile compound with significant applications in electronics,optics, and materials science. It is widely employed in the synthesis of transparentconducting oxides (TCOs), particularly for flat-panel displays, and solarcells, due to its electrical conductivity and optical transparency. Insemiconductor technology, it is used for making indium tin oxide (ITO),enhancing the performance of electronic devices, and is sensitive to variousgases, making it suitable for gas sensing applications, particularly indetecting hazardous gases.

Application

  • Amine Functionalized Surface Frustrated Lewis Pairs for CO2 Photocatalysis: Discusses the enhancement of photocatalytic performance for CO2 reduction using indium oxide hydroxide with amine-functionalized surface frustrated Lewis pairs (Q Guan et al., 2024).
  • Enhancing Gas Sensing Performance through UV Photoexcitation: Explores the improvement of room-temperature gas sensing capabilities of metal oxide semiconductor chemiresistors, including indium(III) oxide, by 400 nm UV photoexcitation (S Paul et al., 2024).
  • Indium(III) Complexes in Industry and Nanoparticle Synthesis: Reviews the use of trivalent indium complexes as catalysts and precursors for various industrial applications and the synthesis of nanoparticles like indium oxide (TO Ajiboye et al., 2024).
  • Ag/In2O3 Inverse Opal Synthesis: Details the synthesis and perspectives of silver/indium oxide inverse opal structures, highlighting their potential in semiconductor applications due to their optical properties (AV Lyutova et al., 2024).
  • Photocatalytic Generation of Hydroxyl Radicals and Manganese Species: Investigates the use of indium oxide in enhancing the photocatalytic performance of permanganate for efficient micropollutant removal under visible light (J Li et al., 2024).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mareike V Hohmann et al.
Journal of physics. Condensed matter : an Institute of Physics journal, 23(33), 334203-334203 (2011-08-05)
The ionization potentials of In(2)O(3) films grown epitaxially by magnetron sputtering on Y-stabilized ZrO(2) substrates with (100) and (111) surface orientation are determined using photoelectron spectroscopy. Epitaxial growth is verified using x-ray diffraction. The observed ionization potentials, which directly affect
Jiefu Yin et al.
Inorganic chemistry, 51(12), 6529-6536 (2012-06-06)
We report here for the first time the hollow, metastable, single-crystal, rhombohedral In(2)O(3) (rh-In(2)O(3)) nanocrystals synthesized by annealing solvothermally prepared InOOH solid nanocrystals under ambient pressure at 400 °C, through a mechanism of the Kirkendall effect, in which pore formation
Di Chen et al.
Nanoscale, 4(10), 3001-3012 (2012-04-13)
With the features of high mobility, a high electric on/off ratio and excellent transparency, metal oxide nanowires are excellent candidates for transparent thin-film transistors, which is one of the key technologies to realize transparent electronics. This article provides a comprehensive
Adrien Mekki-Berrada et al.
Physical chemistry chemical physics : PCCP, 14(12), 4155-4161 (2012-02-16)
The design of new amphoteric catalysts is of great interest for several industrial processes, especially those covering dehydration and dehydrogenation phenomena. Adsorption microcalorimetry was used to monitor the design of mixed oxides of zinc with Group 3 elements (aluminium, gallium
Wen-Ku Chang et al.
Nanomedicine : nanotechnology, biology, and medicine, 8(5), 609-617 (2011-10-29)
Antibacterial activity of photocatalytic substrates is primarily induced by ultraviolet light irradiation. Visible light-responsive photocatalysts were recently discovered, offering greater opportunity to use photocatalysts as disinfectants in our living environment. The development of antibacterial photocatalysts, however, has mainly focused on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service