Skip to Content
Merck
All Photos(1)

Documents

604860

Sigma-Aldrich

L-Glutamic acid-13C5

98 atom % 13C, 95% (CP)

Synonym(s):

(S)-2-Aminopentanedioic acid-13C5, L-Glutamic acid-UL-13C, 13C Labeled L-glutamic acid, Glu-13C5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO213C(13CH2)213CH(NH2)13CO2H
CAS Number:
Molecular Weight:
152.09
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % 13C

Assay

95% (CP)

form

solid

optical activity

[α]25/D +31.0°, c = 2 in 5 M HCl

mp

205 °C (dec.) (lit.)

mass shift

M+5

SMILES string

N[13C@@H]([13CH2][13CH2][13C](O)=O)[13C](O)=O

InChI

1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1/i1+1,2+1,3+1,4+1,5+1

InChI key

WHUUTDBJXJRKMK-WIAREEORSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glutamic acid is an amino acid with an acidic side group. It works as a proton donor. Decarboxylation of glutamic acid results in the formation of GABA (γ aminobutyric acid), a potent neurotransmitter. Isotope labeled amino acids are required for the production of isotopically labeled proteins.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fumihiro Eto et al.
Scientific reports, 10(1), 7435-7435 (2020-05-06)
The brain consists of various areas with anatomical features. Neurons communicate with one another via excitatory or inhibitory synaptic transmission. Altered abundance of neurotransmitters, including glutamate and gamma-aminobutyric acid (GABA), in specific brain regions is closely involved in severe neurological
Garrett RH and Grisham CM
Biochemistry (2012)
The incorporation of radioactive amino acids into proteins of the microsome fraction of guinea-pig liver at very short time intervals after administration.
Simkin JL and Sutton CR
The Biochemical Journal, 77, 489-492 (1960)
Harvey RA, et al.
Biochemistry (Lippincott's illustrated reviews) (2011)
Incorporation of radioactive amino acids into proteins of the microsome fraction of guinea-pig liver in a cell-free system.
SIMKIN JL and WORK TS
The Biochemical Journal, 67, 617-624 (1957)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service