572543
Woollins′ Reagent
Synonym(s):
2,4-Diphenyl-1,3-diselenadiphosphetane 2,4-diselenide
About This Item
Recommended Products
form
solid
mp
206-210 °C (lit.)
storage temp.
2-8°C
SMILES string
[Se]=P1([Se]P(=[Se])([Se]1)c2ccccc2)c3ccccc3
InChI
1S/C12H10P2Se4/c15-13(11-7-3-1-4-8-11)17-14(16,18-13)12-9-5-2-6-10-12/h1-10H
InChI key
GKYFQRQKPYPLGJ-UHFFFAOYSA-N
Related Categories
Application
- Benzoic hydrazides to yield spiro heterocycles
- Secondary amines
- Aromatic diols to yield 10-membered phosphorus-selenium heterocycles
Reagent involved in ring-opening, deselenation and phosphorus-phosphorus coupling
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service