Skip to Content
Merck
All Photos(1)

Key Documents

294799

Sigma-Aldrich

Chloromethane solution

1.0 M in diethyl ether

Synonym(s):

Methyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH3Cl
CAS Number:
Molecular Weight:
50.49
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

concentration

1.0 M in diethyl ether

impurities

<0.005% water

density

0.742 g/mL at 25 °C

functional group

alkyl halide
chloro

storage temp.

2-8°C

SMILES string

CCl

InChI

1S/CH3Cl/c1-2/h1H3

InChI key

NEHMKBQYUWJMIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Radiolysis of aqueous solution of chloromethane was performed with γ-rays. Dechlorination of chloromethane on iron and palladium-iron bimetallic surface in aqueous systems has been reported.

Packaging

Supplied in a Sure/Pac cylinder and has a 1/4-turn bronze ball valve with a female 1/4" NPT outlet thread (Z122661) installed. Before using the cylinder, ensure that the valve is closed, then remove carbon steel plug that seals the outlet valve.

Compatible with the following:For instructions please refer to technical bulletin AL-136: The Aldrich Sure/Pac Cylinder System

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

septum inlet adapter

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 1 - Ozone 1 - Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Dechlorination of chloromethanes on iron and palladium-iron bimetallic surface in aqueous systems.
Wan C, et al
Environmental Toxicology and Chemistry / Setac, 18(6), 1091-1096 (1999)
High dose radiolysis of aqueous solutions of chloromethanes: Importance in the storage of radioactive organic wastes.
Rajesh P, et al.
Journal of Nuclear Materials, 361(1), 10-17 (2007)
Diana Puigserver et al.
Journal of contaminant hydrology, 144(1), 1-19 (2012-11-15)
This study seeks to improve our understanding of the conceptual model of pollutant transport and fate in cases of DNAPL contamination at sites with a complex contamination history. The study was carried out in an unconfined aquifer of alluvial fans
The global chloromethane cycle: biosynthesis, biodegradation and metabolic role.
D B Harper
Natural product reports, 17(4), 337-348 (2000-10-03)
Stéphane Vuilleumier et al.
Journal of bacteriology, 193(18), 5035-5036 (2011-08-27)
Hyphomicrobium sp. strain MC1 is an aerobic methylotroph originally isolated from industrial sewage. This prosthecate bacterium was the first strain reported to grow with chloromethane as the sole carbon and energy source. Its genome, consisting of a single 4.76-Mb chromosome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service