Skip to Content
Merck
All Photos(1)

Documents

563110

Sigma-Aldrich

Diethyl azodicarboxylate solution

40 wt. % in toluene

Synonym(s):

1,2-Ethoxycarbonyl diazene solution, DEAD, Diethoxycarbonyldiazene solution, Diethyl azodiformate solution, NSC 3474, NSC 679015, Unifoam AZ-AE 200

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCON=NCOOC2H5
CAS Number:
Molecular Weight:
174.15
Beilstein:
908662
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

concentration

40 wt. % in toluene

refractive index

n20/D 1.4690

bp

116-117 °C

density

0.956 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CCOC(=O)\N=N/C(=O)OCC

InChI

1S/C6H10N2O4/c1-3-11-5(9)7-8-6(10)12-4-2/h3-4H2,1-2H3/b8-7+

InChI key

FAMRKDQNMBBFBR-BQYQJAHWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DOT Special Approval Submission is Under Review. Availability is Estimated as March 2012.

Application

Reactant for preparation of:
  • Immunostimulants α-Galactosylceramides
  • Cellotriose and cellotetraose analogues as transition state mimics for mechanistic studies of cellulases
  • Bisubstrate inhibitors with molecular recognition at the active site of catechol-O-methyltransferase
  • Derivatives of F200 and S383 with cannabinoid CB1 receptor binding activities
  • Aza-β-lactams via NHC-catalyzed [2 + 2] cycloaddition with ketenes

Reagent for:
  • Annulation of N-protected imines
  • α-thiocyanation of enolizable ketones with ammonium thiocyanate
  • Diels-Alder reactions
A commonly used activating reagent in Mitsunobo reactions.

Packaging

100g unit contains ca. 40g DEAD solute
25g unit contains ca. 10g DEAD solute

Warning

Toxic, Irritant, Flammable

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Repr. 2 - Self-react. C - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

105.8 °F - closed cup

Flash Point(C)

41 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregory S Watson et al.
Acta biomaterialia, 21, 109-122 (2015-03-17)
Geckos, and specifically their feet, have attracted significant attention in recent times with the focus centred around their remarkable adhesional properties. Little attention however has been dedicated to the other remaining regions of the lizard body. In this paper we
A Fürstner et al.
The Journal of organic chemistry, 65(6), 1738-1742 (2000-04-06)
A concise synthesis of the hexahydroazepine moiety 13 of (-)-balanol 1 is described that comprises only eight steps and is distinctly shorter than all previous reported approaches to this particular compound. Sharpless epoxidation of divinylcarbinol 4 and ring closing alkene
Jetze Visser et al.
Nature communications, 6, 6933-6933 (2015-04-29)
Despite intensive research, hydrogels currently available for tissue repair in the musculoskeletal system are unable to meet the mechanical, as well as the biological, requirements for successful outcomes. Here we reinforce soft hydrogels with highly organized, high-porosity microfibre networks that
Basile Siewe et al.
PloS one, 9(4), e92934-e92934 (2014-04-18)
During HIV infection, IL-10/IL-10 receptor and programmed death-1 (PD-1)/programmed death-1-ligand (PD-L1) interactions have been implicated in the impairment of cytotoxic T lymphocyte (CTL) activity. Despite antiretroviral therapy (ART), attenuated anti-HIV CTL functions present a major hurdle towards curative measures requiring
A Ranga et al.
Nature communications, 5, 4324-4324 (2014-07-17)
The behaviour of mammalian cells in a tissue is governed by the three-dimensional (3D) microenvironment and involves a dynamic interplay between biochemical and mechanical signals provided by the extracellular matrix (ECM), cell-cell interactions and soluble factors. The complexity of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service