544302
Methyl 4-acetylbenzoate
98%
Synonym(s):
4-Acetobenzoic acid methyl ester
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3COC6H4CO2CH3
CAS Number:
Molecular Weight:
178.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
98%
mp
93-96 °C (lit.)
functional group
ester
ketone
SMILES string
COC(=O)c1ccc(cc1)C(C)=O
InChI
1S/C10H10O3/c1-7(11)8-3-5-9(6-4-8)10(12)13-2/h3-6H,1-2H3
InChI key
QNTSFZXGLAHYLC-UHFFFAOYSA-N
General description
Methyl 4-acetylbenzoate is an aromatic ketone that can be prepared from 4-bromoacetophenone.
Application
Methyl 4-acetylbenzoate may be used in the preparation of 4-methoxycarbonyl-α-oxo-benzeneacetic acid and methyl 4-(2-(2-methylimidazo[1,2-a]pyridin-3-yl)-2-oxoacetyl)benzoate.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Regioselective Copper?Catalyzed Oxidative Cross?Coupling of Imidazo [1,2?a] pyridines with Methyl Ketones: An Efficient Route for Synthesis of 1,2?Diketones.
Lei S, et al.
Advanced Bacterial Genetics, Cold Spring Harbor, 358(1), 67-73 (2016)
Enantioselective photochemical synthesis of a ?-lactam via the solid state ionic chiral auxiliary method.
Scheffer JR and Wang K.
Synthesis, 112(08), 1253-1257 (2001)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service