Skip to Content
Merck
All Photos(1)

Key Documents

533610

Sigma-Aldrich

2-Acetyl-5-methylthiophene

98%

Synonym(s):

1-(5-Methyl-2-thienyl)ethanone, 1-(5-Methylthiophen-2-yl)ethan-1-one, 1-(5-Methylthiophen-2-yl)ethanone, 2-Methyl-5-acetylthiophene, 5-Methyl-2-thienyl methyl ketone, Methyl 5-methyl-2-thienyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8OS
CAS Number:
Molecular Weight:
140.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.561 (lit.)

bp

65-67 °C/1 mmHg (lit.)

mp

24-28 °C (lit.)

density

1.106 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

CC(=O)c1ccc(C)s1

InChI

1S/C7H8OS/c1-5-3-4-7(9-5)6(2)8/h3-4H,1-2H3

InChI key

YOSDTJYMDAEEAZ-UHFFFAOYSA-N

General description

2-Acetyl-5-methylthiophene is a volatile organic compound formed during the reaction between L-cysteine and dihydroxyacetone in glycerine or triglyceride solvent system. It can be prepared by reacting 2-methylthiophene with acetic anhydride. 2-Acetyl-5-methylthiophene undergoes palladium-catalyzed cross-coupling reaction with aryl bromides to form C-4 arylated product. It reacts with 1,2-bis(5-formyl-2-methylthiophen-3-yl)cyclopentene via Aldol condensation to form a chalcone with photochromic property. The standard molar enthalpies of combustion, formation and vaporization of 2-acetyl-5-methylthiophene are 4341.9 ± 1.8kJ/mol, 158.0 ± 2.1kJ/mol and 62.0 ± 2.6kJ/mol, respectively.

Application

2-Acetyl-5-methylthiophene may be used in the preparation of:
  • 2-ethyl-5-methylthiophene
  • (5-methylthiophen-2-yl)glyoxal
  • (2E)-1-(5-methylthiophen-2-yl)-3-(pyridin-3-yl)prop-2-en-1-one
  • ethyl 3-(5-methylthiophen-2-yl)-3-oxopropanoate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel Dithienylethenes with Extended ??Systems: Synthesis by Aldol Condensation and Photochromic Properties.
Altenhoner K, et al.
European Journal of Organic Chemistry, 2010(31), 6033-6037 (2010)
Palladium-catalysed direct 3-or 4-arylation of thiophene derivatives using aryl bromides.
Dong JJ, et al.
Tetrahedron, 50(23), 2778-2781 (2009)
Synthesis, Configuration, and Antimicrobial Properties of Novel Substituted and Cyclized 2?, 3??-Thiazachalcones.
Ustaa A, et al.
Helvetica Chimica Acta, 90, 1482-1482 (2007)
Acylation studies in the thiophene and furan series. IV. Strong inorganic oxyacids as catalysts.
Hartough HD and Kosak AI.
Journal of the American Chemical Society, 69(12), 3093-3096 (1947)
Experimental thermochemical study of the three methyl substituted 2-acetylthiophene isomers.
da Silva MAVR and Santos AFLOM.
The Journal of Chemical Thermodynamics, 40(8), 1309-1313 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service