Skip to Content
Merck
All Photos(1)

Documents

288411

Sigma-Aldrich

2-Acetylthiazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5NOS
CAS Number:
Molecular Weight:
127.16
Beilstein:
109803
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

refractive index

n20/D 1.548 (lit.)

bp

89-91 °C/12 mmHg (lit.)

density

1.227 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1nccs1

InChI

1S/C5H5NOS/c1-4(7)5-6-2-3-8-5/h2-3H,1H3

InChI key

MOMFXATYAINJML-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Acetylthiazole is one of the components of wine and was quantified by GC-MS. It is also found in volatile oil from Boletopsis leucomelas (Pers.) Fayod.

Application

2-Acetylthiazole was used in the preparation of triazolothiazoles, chiral alcohols and in aldol condensation reactions.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sota Nosaka et al.
Journal of oleo science, 63(6), 577-583 (2014-06-03)
The volatile oil from Boletopsis leucomelas (Pers.) Fayod was extracted by hydrodistillation with diethylether, and the volatile components of the oil were analyzed by gas chromatography-mass spectrometry. The oil contained 86 components, representing 87.5% of the total oil. The main
Tetrahedron, 47, 2851-2851 (1991)
Tetrahedron Asymmetry, 2, 243-243 (1991)
The Journal of Organic Chemistry, 56, 5294-5294 (1991)
Harald Keim et al.
Journal of agricultural and food chemistry, 50(21), 5803-5807 (2002-10-03)
Qualitative and quantitative analyses of N-heterocycle compounds were conducted by using a liquid-liquid extraction followed by additional chemical silica gel purification and injection into a gas chromatographic (GC) column coupled to a nitrogen-phosphorus detector (NPD). The purified extract fraction was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service