Skip to Content
Merck
All Photos(2)

Key Documents

513431

Sigma-Aldrich

Luperox® AFR40, Benzoyl peroxide solution

40 wt. % in dibutyl phthalate

Synonym(s):

Benzoyl peroxide, blend in dibutyl phthalate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5CO)2O2
CAS Number:
Molecular Weight:
242.23
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

reaction suitability

reagent type: oxidant

concentration

40 wt. % in dibutyl phthalate

storage temp.

2-8°C

SMILES string

O=C(OOC(=O)c1ccccc1)c2ccccc2

InChI

1S/C14H10O4/c15-13(11-7-3-1-4-8-11)17-18-14(16)12-9-5-2-6-10-12/h1-10H

InChI key

OMPJBNCRMGITSC-UHFFFAOYSA-N

Application

Polymerization initiator.

Legal Information

Product of Arkema Inc.
Luperox is a registered trademark of Arkema Inc.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Org. Perox. E - Repr. 1B - Skin Sens. 1

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emil A Tanghetti et al.
Dermatologic clinics, 27(1), 17-24 (2008-11-06)
Benzoyl peroxide (BPO) is the most widely used topical acne treatment, with significant antibacterial, antikeratolytic, and comedolytic activity. It has been shown to be extremely effective as monotherapy and in combination with antibiotics or retinoids for managing comedonal and inflammatory
Suzanne M Jervis et al.
Journal of food science, 78(2), R129-R137 (2013-01-22)
Whey is a value-added product that is utilized in many food and beverage applications for its nutritional and functional properties. Whey and whey products are generally utilized in dried ingredient applications. One of the primary sources of whey is from
Adam J Friedman et al.
The Journal of investigative dermatology, 133(5), 1231-1239 (2012-11-30)
Advances in nanotechnology have demonstrated potential application of nanoparticles (NPs) for effective and targeted drug delivery. Here we investigated the antimicrobial and immunological properties and the feasibility of using NPs to deliver antimicrobial agents to treat a cutaneous pathogen. NPs
Lawrence F Eichenfield et al.
Journal of drugs in dermatology : JDD, 11(7), 818-824 (2012-07-11)
Acne is common in adolescents and especially difficult to manage in people with color. A fixed combination of clindamycin phosphate and benzoyl peroxide (BPO) (clindamycin phosphate 1.2%/BPO 2.5% gel) was evaluated to determine its utility in treating moderate to severe
E J Kang et al.
Journal of food science, 77(7), C818-C823 (2012-07-05)
Residual annatto colorant (norbixin) in fluid Cheddar cheese whey can be bleached. The 2 approved chemical bleaching agents for whey, hydrogen peroxide (HP) and benzoyl peroxide (BP), negatively impact the flavor of dried whey protein. The objective of this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service