Skip to Content
Merck
All Photos(3)

Key Documents

E9508

Sigma-Aldrich

Ethanolamine

≥98%

Synonym(s):

2-Aminoethanol, 2-Aminoethyl alcohol, ETA, MEA, MEA 90, MEA-LCI, Monoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2OH
CAS Number:
Molecular Weight:
61.08
Beilstein:
505944
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

vapor density

2.1 (vs air)

Quality Level

vapor pressure

0.2 mmHg ( 20 °C)

Assay

≥98%

form

liquid

autoignition temp.

1436 °F

expl. lim.

17 %

refractive index

n20/D 1.454 (lit.)

pH

12.1 (20 °C, 100 g/L)

pKa (25 °C)

9.5(lit.)

bp

170 °C (lit.)
69-70 °C/10 mmHg

mp

10-11 °C (lit.)

density

1.012 g/mL at 25 °C (lit.)

SMILES string

NCCO

InChI

1S/C2H7NO/c3-1-2-4/h4H,1-3H2

InChI key

HZAXFHJVJLSVMW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • New Functionalized Di-substituent Imidazolium Ionic Liquids as Superior Faster Absorbents for Carbon Dioxide Gas.: This research highlights the development of novel imidazolium ionic liquids, where ethanolamine plays a crucial role in enhancing their efficiency as fast absorbents for carbon dioxide removal, presenting a potential impactful application in environmental science and engineering (Shawish et al., 2024).

  • Role of ethanolamine utilization and bacterial microcompartment formation in Listeria monocytogenes intracellular infection.: This study explores how Listeria monocytogenes utilizes ethanolamine and forms bacterial microcompartments, offering insights into bacterial pathogenesis and potential targets for antibiotic development (Chatterjee et al., 2024).

  • (13)C-Stable isotope resolved metabolomics uncovers dynamic biochemical landscape of gut microbiome-host organ communications in mice.: Research here utilizes ethanolamine in stable isotope-resolved metabolomics to map the dynamic biochemical interactions between the gut microbiome and host organs, advancing our understanding of microbial contributions to host metabolism (Xiao et al., 2024).

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

195.8 °F - Pensky-Martens closed cup

Flash Point(C)

91 °C - Pensky-Martens closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wen-Chih Cheng et al.
Nature communications, 11(1), 4774-4774 (2020-09-24)
Detection of microbial nucleic acids in body fluids has become the preferred method for rapid diagnosis of many infectious diseases. However, culture-based diagnostics that are time-consuming remain the gold standard approach in certain cases, such as sepsis. New culture-free methods
Yacine Bounab et al.
Nature protocols, 15(9), 2920-2955 (2020-08-14)
Characterization of immune responses is currently hampered by the lack of systems enabling quantitative and dynamic phenotypic characterization of individual cells and, in particular, analysis of secreted proteins such as cytokines and antibodies. We recently developed a simple and robust
Michael Backlund et al.
Nucleic acids research, 48(9), 4725-4740 (2020-04-22)
Cellular stress causes multifaceted reactions to trigger adaptive responses to environmental cues at all levels of the gene expression pathway. RNA-binding proteins (RBP) are key contributors to stress-induced regulation of RNA fate and function. Here, we uncover the plasticity of
Florent Arbogast et al.
Autophagy, 15(2), 280-294 (2018-09-11)
The involvement of macroautophagy/autophagy proteins in B-cell receptor (BCR) trafficking, although suspected, is not well understood. We show that ATG5 (autophagy related 5) contributes to BCR polarization after stimulation and internalization into LAMP1 (lysosomal-associated membrane protein 1)+ and major histocompatibility
Gabriel da Silva
The journal of physical chemistry. A, 116(45), 10980-10986 (2012-09-22)
The alkanolamine 2-aminoethanol (NH(2)CH(2)CH(2)OH), otherwise known as monoethanolamine (MEA), is a widely used solvent for carbon capture, yet relatively little is known about its atmospheric chemistry. The hydroxyl radical initiated oxidation of MEA is thought to predominantly form the α-aminoalkyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service