Skip to Content
Merck
All Photos(4)

Key Documents

497401

Sigma-Aldrich

(R)-(+)-2-Methyl-2-propanesulfinamide

98%

Synonym(s):

(R)-2-methyl-2-propanesulfinamide, (R)-2-methylpropane-2-sulfinamide, (R)-tert-butanesulfinamide, (R)-tert-butylsulfinamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CS(O)NH2
CAS Number:
Molecular Weight:
121.20
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

optical activity

[α]20/D +4°, c = 1.0242 in chloroform stab. with amylenes

mp

103-107 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)S(N)=O

InChI

1S/C4H11NOS/c1-4(2,3)7(5)6/h5H2,1-3H3/t7-/m1/s1

InChI key

CESUXLKAADQNTB-SSDOTTSWSA-N

General description

(R)-(+)-2-Methyl-2-propanesulfinamide is a chiral auxiliary used in the condensation of the aldehyde.

Application

(R)-(+)-2-Methyl-2-propanesulfinamide may be used to prepare N-(1-cyclohexylmethylidene)-2-methylpropane-2-sulfinamide via copper mediated condensation with cyclohexane carboxaldehyde. It may also be used to prepare (20E)-N-[t-butyl-(R)-sulfinyl]-3β-(t-butyldimethylsilyloxy)-pregn-5-en-20-imine, an intermediate for the development of androgen receptor antagonists.
Can be readily transformed into P,N-sulfinyl imine ligands through condensation with aldehydes and ketones, which can undergo iridium-catalyzed asymmetric hydrogenation of olefins.
Preparation of ß-chloro sulfinamides in a synthesis of chiral azridines. Also used to prepare an organocatalyst for enantioselective reduction of imines.
Useful reagent for synthesizing chiral amines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bram Denolf et al.
Organic letters, 8(14), 3129-3132 (2006-06-30)
[reaction: see text] Reaction of chiral alpha-chloro tert-butanesulfinyl aldimines with Grignard reagents efficiently afforded beta-chloro N-sulfinamides in high diastereomeric excess. The latter compounds were cyclized toward the corresponding chiral aziridines in a high-yielding one-pot reaction or after separate treatment with
20-Aminosteroids as a novel class of selective and complete androgen receptor antagonists and inhibitors of prostate cancer cell growth.
Fousteris MA, et al.
Bioorganic & Medicinal Chemistry, 18(19), 6960-6969 (2010)
Jeffrey P McMahon et al.
Organic letters, 6(10), 1645-1647 (2004-05-07)
Addition of alkyl or aryl Grignard reagents to N-sulfinyl imines derived from 3- and 4-substituted cyclohexanones proceeds with good yields and with excellent diasteroselectivity. The selectivity of the reaction is controlled by the ring substituent rather than the sulfinyl group
Asymmetric synthesis of a, a-difluoro-?-amino acid derivatives from enantiomerically pure N-tert-butylsulfinimines.
Staas, DD, et al.
The Journal of Organic Chemistry, 67(23), 8276-8279 (2002)
Dong Pei et al.
Organic letters, 8(25), 5913-5915 (2006-12-01)
Easily accessible chiral sulfinamide 2 has been developed as the first highly efficient and enantioselective organocatalyst relying solely on a chiral sulfur center for stereochemical induction. In the presence of 20 mol % of 2, a broad range of N-aryl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service