Skip to Content
Merck
All Photos(1)

Documents

382183

Sigma-Aldrich

Benzyloxyacetaldehyde

97%

Synonym(s):

α-(Benzyloxy)acetaldehyde, (Benzyloxy)acetaldehyde, (Benzyloxy)ethanal, (Phenylmethoxy)acetaldehyde, 2-(Benzyloxy)acetaldehyde, 2-(Benzyloxy)ethanal, 2-(Phenylmethoxy)acetaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OCH2CHO
CAS Number:
Molecular Weight:
150.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

contains

0.5% hydroquinone as stabilizer

refractive index

n20/D 1.518 (lit.)

bp

118-120 °C/13 mmHg (lit.)

density

1.069 g/mL at 25 °C (lit.)

SMILES string

O=CCOCc1ccccc1

InChI

1S/C9H10O2/c10-6-7-11-8-9-4-2-1-3-5-9/h1-6H,7-8H2

InChI key

NFNOAHXEQXMCGT-UHFFFAOYSA-N

General description

Benzyloxyacetaldehyde is a non-natural aldehyde. It undergoes enantioselective Mukaiyama aldol reaction with silylketene acetal nucleophiles in the presence of C2-symmetric bis(oxazolinyl)pyridine Cu(II) complex (catalyst).

Application

Benzyloxyacetaldehyde may be used in the following syntheses:
  • (3S,5S)-methyl 6-benzyloxy-3,5-dihydroxyhexanoate
  • (S)-5-benzyloxy-4-hydroxypentan-2-one
  • myxothiazols
Benzyloxyacetaldehyde may be used in the synthesis of 1-deoxy-L-xylulose, via antibody catalysis.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Vidal et al.
Applied microbiology and biotechnology, 68(4), 489-497 (2005-02-24)
The glyA gene encoding a serine hydroxymethyl transferase (SHMT) with threonine aldolase activity was isolated from Streptococcus thermophilus YKA-184 chromosomal DNA. This aldolase is a pyridoxal 5'-phosphate-dependent enzyme that stereospecifically catalyzes the interconversion of L-threonine to glycine and acetaldehyde. The
A short enantioselective synthesis of 1-deoxy-L-xylulose by antibody catalysis.
Shabat D, et al.
Tetrahedron Letters, 40(8), 1437-1440 (1999)
C2-symmetric copper (II) complexes as chiral Lewis acids. Scope and mechanism of catalytic enantioselective aldol additions of enolsilanes to (benzyloxy) acetaldehyde.
Evans DA, et al.
Journal of the American Chemical Society, 121(4), 669-685 (1999)
Evans DA, et al.
Journal of the American Chemical Society, 121, 669-685 (1999)
Formal total syntheses of Myxothiazols by three different approaches starting from benzyloxyacetaldehyde.
Backhaus D.
Tetrahedron Letters, 41(13), 2087-2090 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service