Skip to Content
Merck
All Photos(1)

Documents

495239

Sigma-Aldrich

[(1,1-Dimethyl-2-propynyl)oxy]trimethylsilane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CC(CH3)2OSi(CH3)3
CAS Number:
Molecular Weight:
156.30
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

refractive index

n20/D 1.404 (lit.)

bp

115-116 °C (lit.)

density

0.806 g/mL at 25 °C (lit.)

SMILES string

CC(C)(O[Si](C)(C)C)C#C

InChI

1S/C8H16OSi/c1-7-8(2,3)9-10(4,5)6/h1H,2-6H3

InChI key

JNRUXZIXAXHXTN-UHFFFAOYSA-N

General description

[(1,1-Dimethyl-2-propynyl)oxy]trimethylsilane is also known as trimethyl[(2-methyl-3-butyn-2-yl)oxy]silane. Its enthalpy of vaporization at boiling point has been reported.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

68.0 °F

Flash Point(C)

20 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaws CL.
Thermophysical Properties of Chemicals and Hydrocarbons, 560-560 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service