Skip to Content
Merck
All Photos(1)

Key Documents

A32608

Sigma-Aldrich

Allyl glycidyl ether

≥99%

Synonym(s):

1-Allyloxy-2,3-epoxypropane, Allyl 2,3-epoxypropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
Beilstein:
105871
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor density

3.9 (vs air)

Quality Level

vapor pressure

4.7 mmHg ( 25 °C)

Assay

≥99%

form

liquid

refractive index

n20/D 1.433 (lit.)

bp

154 °C (lit.)

density

0.962 g/mL at 25 °C (lit.)

SMILES string

C=CCOCC1OC1

InChI

1S/C6H10O2/c1-2-3-7-4-6-5-8-6/h2,6H,1,3-5H2

InChI key

LSWYGACWGAICNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Allyl glycidyl ether (AGE) is a monomer that acts as a building block in the polymerization of poly(allyl glycidyl ether) by anionic ring opening mechanism. It is industrially prepared by the reacting allyl alcohol and epichlorohydrin.[1][2]

Application

AGE can be used as a terminal epoxy group in the development of adhesive surfaces by hydrosilylation process.[3] It can also be used in the synthesis of polymeric electrolytes for the fabrication of lithium batteries.[4]
Useful in hydrosilylation, including the H2PtCl6·xH2O (cat. no. 206083) catalyzed hydrosilylation of siloxanes to epoxysiloxanes.[5]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F - closed cup

Flash Point(C)

57 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minimally adhesive polymer surfaces prepared from star oligosiloxanes and star oligofluorosiloxanes
Grunlan MA, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 44(8), 2551-2566 (2006)
Y Koyama et al.
Bioconjugate chemistry, 7(3), 298-301 (1996-05-01)
Novel poly(ethylene glycol) (PEG) derivatives having pendant amino groups were prepared by copolymerization of allyl glycidyl ether with ethylene oxide followed by chemical modification of the double bond side chains. Dropwise addition of the mixture of monomers to the anionic
Bhalchandra K Vaidya et al.
Bioresource technology, 99(9), 3623-3629 (2007-09-04)
Macroporous polymer particles containing surface epoxy groups were synthesized for immobilization of Candida rugosa lipase (CRL). The effect of incorporation of two different sets of monomers [allyl glycidyl ether (AGE) and glycidyl methacrylate (GMA)] and the effect of crosslinking density
Challa Lahari et al.
Langmuir : the ACS journal of surfaces and colloids, 26(2), 1096-1106 (2009-12-10)
Effects of changes in hydrophobicity of polymeric support on structure and activity of alpha-chymotrypsin (E.C. 3.4.21.1) have been studied with copolymers of allyl glycidyl ether (AGE) and ethylene glycol dimethacrylate (EGDM) with increasing molar ratio of EGDM to AGE (cross-link
Allyl glycidyl ether-based polymer electrolytes for room temperature lithium batteries
Barteau KP, et al.
Macromolecules, 46(22), 8988-8994 (2013)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service