Skip to Content
Merck
All Photos(1)

Key Documents

474754

Sigma-Aldrich

Magnesium

dendritic pieces, purified by distillation, 99.998% trace metals basis

Synonym(s):

Hypermagnesemia, Magnesium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
Beilstein:
4948473
EC Number:
MDL number:
UNSPSC Code:
12141502
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 621 °C)

Quality Level

Assay

99.998% trace metals basis

form

dendritic pieces

autoignition temp.

950 °F

purified by

distillation

reaction suitability

reagent type: reductant

resistivity

4.46 μΩ-cm, 20°C

bp

1090 °C (lit.)

mp

648 °C (lit.)

density

1.74 g/mL at 25 °C (lit.)

SMILES string

[Mg]

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Solid state synthesis with Ca and Sn resulted in a new phase, Ca6.2Mg3.8Sn7, which has an unprecedented type of tin chain composed of square-planar tin units.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Sol. 1 - Water-react 1

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ganguli, A.K. et al.
Journal of the American Chemical Society, 120, 1223-1223 (1998)
Shuvendu Biswas et al.
Nature chemistry, 5(7), 613-620 (2013-06-22)
The development of nanocarriers that selectively release guest molecules on sensing a particular biological signal is being actively pursued in nanomedicine for diagnostic and therapeutic purposes. Here we report a protein-based nanocarrier that opens in the presence of intracellular adenosine-5'-triphosphate
Benjamin Chaigne-Delalande et al.
Science (New York, N.Y.), 341(6142), 186-191 (2013-07-13)
The magnesium transporter 1 (MAGT1) is a critical regulator of basal intracellular free magnesium (Mg(2+)) concentrations. Individuals with genetic deficiencies in MAGT1 have high levels of Epstein-Barr virus (EBV) and a predisposition to lymphoma. We show that decreased intracellular free
Denise H M Heppe et al.
The American journal of clinical nutrition, 98(1), 224-232 (2013-05-31)
Maternal diet during pregnancy has been suggested to influence bone health in later life. We assessed the association of maternal first-trimester dietary intake during pregnancy with childhood bone mass. In a prospective cohort study in 2819 mothers and their children
Carlos Zarate et al.
Annals of the New York Academy of Sciences, 1292, 21-31 (2013-07-24)
Clinical depression is a serious mental disorder characterized by low mood, anhedonia, loss of interest in daily activities, and other symptoms, and is associated with severe consequences including suicide and increased risk of cardiovascular events. Depression affects nearly 15% of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service