Skip to Content
Merck
All Photos(1)

Key Documents

63035

Sigma-Aldrich

Magnesium

purum, for Grignard reactions, ≥99.5%, turnings

Synonym(s):

Magnesium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Mg
CAS Number:
Molecular Weight:
24.31
Beilstein:
4948473
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

1 mmHg ( 621 °C)

grade

purum

Assay

≥99.5%

form

turnings

autoignition temp.

950 °F

reaction suitability

reaction type: Grignard Reaction
reagent type: reductant

resistivity

4.46 μΩ-cm, 20°C

bp

1090 °C (lit.)

mp

648 °C (lit.)

density

1.74 g/mL at 25 °C (lit.)

SMILES string

[Mg]

InChI

1S/Mg

InChI key

FYYHWMGAXLPEAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Magnesium turnings are mainly used to prepare Grignard reagent to synthesize a wide range of reaction intermediates.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Self-heat. 1 - Water-react 2

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectroscopic and electrical characterization of ?, ?-bisdiphenylene-?-phenylallyl radical as an organic semiconductor.
Matsui Y
Research on Chemical Intermediates, 1-10 (2018)
Medium band gap conjugated polymers from thienoacene derivatives and pentacyclic aromatic lactam as promising alternatives of poly (3?hexylthiophene) in photovoltaic application.
Gao P
Journal of Polymer Science Part A: Polymer Chemistry, 56(1), 85-95 (2018)
Convenient synthesis of perfluoroalkyltrifluoroborates.
Sprenger JA
Journal of Fluorine Chemistry, 206, 54-60 (2018)
Shuvendu Biswas et al.
Nature chemistry, 5(7), 613-620 (2013-06-22)
The development of nanocarriers that selectively release guest molecules on sensing a particular biological signal is being actively pursued in nanomedicine for diagnostic and therapeutic purposes. Here we report a protein-based nanocarrier that opens in the presence of intracellular adenosine-5'-triphosphate
Benjamin Chaigne-Delalande et al.
Science (New York, N.Y.), 341(6142), 186-191 (2013-07-13)
The magnesium transporter 1 (MAGT1) is a critical regulator of basal intracellular free magnesium (Mg(2+)) concentrations. Individuals with genetic deficiencies in MAGT1 have high levels of Epstein-Barr virus (EBV) and a predisposition to lymphoma. We show that decreased intracellular free

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service