Skip to Content
Merck
All Photos(1)

Key Documents

468495

Sigma-Aldrich

1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate

97%

Synonym(s):

DiI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C59H97ClN2O4
CAS Number:
Molecular Weight:
933.87
Beilstein:
8251870
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

97%

form

crystalline

composition

Carbon: 73.2-78.5%

Nitrogen: 2.7-3.3%

technique(s)

titration: suitable

color

dark purple

mp

68 °C (dec.) (lit.)

λmax

549 nm

ε (extinction coefficient)

≥10000 at 276-282 nm in methanol at 0.01 g/L
≥130000 at 546-552 nm in methanol at 0.01 g/L
≥70000 at 514-520 nm in methanol at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[O-]Cl(=O)(=O)=O.CCCCCCCCCCCCCCCCCCN1c2ccccc2C(C)(C)/C1=C\C=C\C3=[N+](CCCCCCCCCCCCCCCCCC)c4ccccc4C3(C)C

InChI

1S/C59H97N2.ClHO4/c1-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-41-50-60-54-46-39-37-44-52(54)58(3,4)56(60)48-43-49-57-59(5,6)53-45-38-40-47-55(53)61(57)51-42-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-2;2-1(3,4)5/h37-40,43-49H,7-36,41-42,50-51H2,1-6H3;(H,2,3,4,5)/q+1;/p-1

InChI key

JVXZRNYCRFIEGV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate is also called as Dil. It can be used to study HCV (Hepatitis C virus) cell interactions in the HCV entry process. It is used to label oxidized LDL (low-density lipoprotein) to quantify the uptake of DiI-oxLDL in human endothelial cells.

Application

1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate has been used in cytoskeletal staining, nuclear staining, retrograde labeling, counting of motor neurons and labelling of the retinal ganglion cell axons.
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine perchlorate has been used in fluorescent labelling of mesenchymal stem cells (MSCs) and cerebral cortex. It has also been used in electromyogram (EMG) to determine awake and anesthetized states in mice.

Biochem/physiol Actions

Lipophilic carbocyanine dye used primarily for optical recordings of membrane voltage and for studies of membrane fluidity. Retrograde stain for neurons; provides intense, long-lasting staining of live neurons in vivo and in vitro.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flexible Wireless Powered Drug Delivery System for Targeted Administration on Cerebral Cortex
Sung SH, et al.
Nano Energy (2018)
Angiotensin II induces LOX-1, the human endothelial receptor for oxidized low-density lipoprotein.
Morawietz H, et al.
Circulation, 100(9), 899-902 (1999)
Zaborszky L
Neuroanatomical Tract-Tracing: Molecules, Neurons, and Systems (2006)
Activation of the hypothalamic feeding centre upon visual prey detection.
Muto A, et al.
Nature Communications, 8:15029 (2017)
Bartenschlager R, et al.
Hepatitis C Virus: From Molecular Virology to Antiviral Therapy, 369 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service