467855
Copper(II) nitrate hemi(pentahydrate)
ACS reagent, ≥99.99% trace metals basis
Synonym(s):
Cupric nitrate hemi(pentahydrate)
About This Item
Recommended Products
grade
ACS reagent
Quality Level
Assay
≥99.99% trace metals basis
form
crystalline
impurities
≤0.01% insolubles
<100 ppm total metallic impurities
anion traces
chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.01%
cation traces
Ca: ≤0.005%
Fe: ≤0.005%
K: ≤0.005%
Na: ≤0.01%
Ni: ≤0.01%
Pb: ≤0.001%
application(s)
battery manufacturing
SMILES string
O.O.O.O.O.[Cu++].[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
InChI
1S/2Cu.4NO3.5H2O/c;;4*2-1(3)4;;;;;/h;;;;;;5*1H2/q2*+2;4*-1;;;;;
InChI key
IZDJJEMZQZQQQQ-UHFFFAOYSA-N
Related Categories
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 1 - Skin Corr. 1B
Storage Class Code
5.1A - Strongly oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service