Skip to Content
Merck
All Photos(1)

Documents

443999

Sigma-Aldrich

Trimethyl borate

purified by redistillation, ≥99.5%

Synonym(s):

Boric acid trimethyl ester, Methyl borate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
B(OCH3)3
CAS Number:
Molecular Weight:
103.91
Beilstein:
1697939
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.59 (vs air)

Assay

≥99.5%

form

liquid

purified by

redistillation

refractive index

n20/D 1.346 (lit.)

bp

68-69 °C (lit.)

mp

−34 °C (lit.)

density

0.932 g/mL at 20 °C (lit.)

SMILES string

COB(OC)OC

InChI

1S/C3H9BO3/c1-5-4(6-2)7-3/h1-3H3

InChI key

WRECIMRULFAWHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Trimethyl borate can be used as:
  • A reagent in the preparation of various trialkylamine−boranes from corresponding trialkylamines using lithium hydride/aluminum chloride catalyst.
  • A source of boron in the synthesis of boron nitride (BN) nanotubes by thermal-heating chemical vapor deposition (TH-CVD) method.
  • An electrolytic additive for electrochemical applications.
  • A reagent along with lithium di-tert-butyl(2,2,6,6-tetramethylpiperidino)zincate (TMP-zincate) for the synthesis of 1-tert-butyl-3,4-dihydroisoquinoline from isoquinoline via ortho metalation reaction.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

12.2 °F - (own results)

Flash Point(C)

-11 °C - (own results)

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Trimethyl borate-induced one-pot homologation reactions of isoquinoline with di-tert-butyl-TMP zincate
Seo HJ, et al.
Tetrahedron Letters, 52(29), 3747-3750 (2011)
Trimethyl borate as an electrolyte additive for high potential layered cathode with concurrent improvement of rate capability and cyclic stability
Wang Z, et al.
Electrochimica Acta, 184(1), 40-46 (2015)
Thermal-heating CVD synthesis of BN nanotubes from trimethyl borate and nitrogen gas.
Lin FH, et al.
Materials Chemistry and Physics, 107(1), 115-121 (2008)
One-Pot Synthesis of Ammonia-Borane and Trialkylamine-Boranes from Trimethyl Borate.
Veeraraghavan RP, et al.
Organic Letters, 14(24), 6119-6121 (2012)
R Meder et al.
Solid state nuclear magnetic resonance, 15(1), 69-72 (2000-07-21)
Boron-11 nuclear magnetic resonance imaging and spectroscopy have been used to characterise the nature and distribution of boron compounds after preservative treatment of radiata pine wood with trimethylborate (TMB). One day after treatment, 11B magnetic resonance imaging microscopy showed significant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service