790877
Tris(2,2,2-trifluoroethyl) borate
97%
Synonym(s):
Tris(2,2,2-trifluoroethyl)borate, Sheppard Amidation Reagent, Tris(2,2,2-trifluoroethoxy)boron, Sheppard Reagent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H6BF9O3
CAS Number:
Molecular Weight:
307.91
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
97%
form
liquid
refractive index
n/D 1.298 (lit.)
density
1.430 g/mL at 25 °C
functional group
fluoro
SMILES string
FC(F)(F)COB(OCC(F)(F)F)OCC(F)(F)F
InChI
1S/C6H6BF9O3/c8-4(9,10)1-17-7(18-2-5(11,12)13)19-3-6(14,15)16/h1-3H2
InChI key
DIEXQJFSUBBIRP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
Reagent can promote the direct formation of amides from carboxylic acids and amines under thermal and microwave conditions.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
109.9 °F
Flash Point(C)
43.3 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Rachel M Lanigan et al.
The Journal of organic chemistry, 78(9), 4512-4523 (2013-04-17)
B(OCH2CF3)3, prepared from readily available B2O3 and 2,2,2-trifluoroethanol, is as an effective reagent for the direct amidation of a variety of carboxylic acids with a broad range of amines. In most cases, the amide products can be purified by a
Pavel Starkov et al.
Organic & biomolecular chemistry, 9(5), 1320-1323 (2011-01-08)
Simple borates serve as effective promoters for amide bond formation with a variety of carboxylic acids and amines. With trimethyl or tris(2,2,2-trifluoroethyl) borate, amides are obtained in good to excellent yield and high purity after a simple work-up procedure. Tris(2,2,2-trifluoroethyl)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service