Skip to Content
Merck
All Photos(3)

Documents

430676

Sigma-Aldrich

2,2,5,7,8-Pentamethyl-6-chromanol

97%

Synonym(s):

α-C-1-Chromanol, 2,2,5,7,8-Pentamethyl-3,4-dihydrochromen-6-ol, 2,2,5,7,8-Pentamethyl-6-hydroxychroman, 3,4-Dihydro-2,2,5,7,8-pentamethyl-2H-1-benzopyran-6-ol, 6-Hydroxy-2,2,5,7,8-pentamethylchroman

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20O2
CAS Number:
Molecular Weight:
220.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

89-91 °C (lit.)

SMILES string

Cc1c(C)c2OC(C)(C)CCc2c(C)c1O

InChI

1S/C14H20O2/c1-8-9(2)13-11(10(3)12(8)15)6-7-14(4,5)16-13/h15H,6-7H2,1-5H3

InChI key

SEBPXHSZHLFWRL-UHFFFAOYSA-N

Gene Information

human ... AR(367)

General description

2,2,5,7,8-Pentamethyl-6-chromanol has been reported as an vitamin E model compound. It undergoes oxidation in presence of various alcohols, ranging from methanol to cholesterol, affords 5-alkoxymethyl-2,2,7,8-tetramethyl-6-chromanols. Antioxidant moiety of vitamin E, 2,2,5,7,8-pentamethyl-6-chromanol (PMCol), is reported to have antiandrogen activity in prostate carcinoma cells.

Application

2,2,5,7,8-Pentamethyl-6-chromanol is the suitable reagent used for the quantitative analysis of α-tocopherol by plasma-based gas chromatography/tandem mass spectrometry (GC/MS/MS) using a tabletop quadrupole ion trap mass spectrometer. It may be employed as α-tocopherol model compound and on oxidation by t-butyl hydroperoxide in chloroform, in the presence of alcohol, affords 5-alkoxymethyl-2,2,7,8-tetramethyl-6-chromanol. It may be used as internal standard for the determination of α- and γ-tocopherol in the rabbit serum and liver by HPLC.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julien Chapelat et al.
Organic letters, 10(22), 5123-5126 (2008-10-22)
A diastereoselective synthesis of alpha-tocopherol 1 (93% de) was achieved via two key steps, (i) a highly diastereoselective Shi epoxidation of a trisubstituted alkene and (ii) an acid supported, "anti-Baldwin" epoxide ring opening under inversion of configuration leading to the
M H Kang et al.
The Journal of nutrition, 129(10), 1885-1890 (1999-09-28)
Plant glucosides possess antioxidative properties due to their ability to scavenge free radicals. Sesame seeds contain a class of these compounds, the sesaminol glucosides. To evaluate their antioxidative activity in vivo, we fed rabbits diets containing 1% cholesterol (Chol) with
Katerina Krumova et al.
Journal of the American Chemical Society, 134(24), 10102-10113 (2012-05-10)
The preparation of two highly sensitive fluorogenic α-tocopherol (TOH) analogues which undergo >30-fold fluorescence intensity enhancement upon reaction with peroxyl radicals is reported. The probes consist of a chromanol moiety coupled to the meso position of a BODIPY fluorophore, where
Jolanta Gruszka et al.
Free radical biology & medicine, 45(6), 920-928 (2008-07-19)
Singlet oxygen quenching rate constants for tocopherol and tocotrienol homologues have been determined in organic solvents of different polarities, as well as for other biological prenyllipids such as plastoquinol, ubiquinol, and alpha-tocopherolquinol. The obtained results showed that the quenching activity
Raymond Hreiche et al.
Journal of molecular and cellular cardiology, 47(2), 210-220 (2009-02-24)
Drug-induced QT interval prolongation is a condition likely to be aggravated by diabetes. The objective of this study was to evaluate how glucose concentration may modulate drug effects on ventricular repolarization and on cardiac repolarizing potassium currents. Guinea pig hearts

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service