Skip to Content
Merck
All Photos(1)

Key Documents

06423

Supelco

Amberlyst® 15 Ion Exchange Resin

greener alternative

hydrogen form, >50 mesh, strongly acidic

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52

Product Name

Amberlyst® 15 hydrogen form, strongly acidic, cation exchanger, dry

form

beads

quality

dry

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

parameter

120 °C max. temp.

moisture

≤1.6%

technique(s)

LPLC: suitable

loss

~5% loss on drying, 110 °C

matrix

styrene-divinylbenzene (macroreticular)

matrix active group

sulfonic acid functional group

particle size

<300 μm

operating pH

0-14

capacity

1.7 meq/mL by wetted bed volume
4.7 meq/g by dry weight

greener alternative category

separation technique

cation exchange

InChI

1S/C10H10.C8H8O3S/c1-3-9-7-5-6-8-10(9)4-2;1-2-7-5-3-4-6-8(7)12(9,10)11/h3-8H,1-2H2;2-6H,1H2,(H,9,10,11)

InChI key

SIWVGXQOXWGJCI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Amberlyst-15 is a Brønsted acidic resin catalyst.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

Amberlyst-15 has been used as a catalyst for the etherification of furanyl alcohols by using both ethanol and butanol solutions. Amberlyst-15 hydrogen form has been used as an acid resin catalyst for the conversion of furfuryl alcohol into ethyl levulinate in lipid ethanol and for biodiesel production from triolein in a batch reaction system by transesterification method at high temperatures.
Strongly acidic cation exchange resin used as a heterogeneous acid catalysis; suitable for non-aqueous catalysis.

Chem/Phys Resistance

Swelling (dry to solvent saturated, %):
hexane 10-15; toluene 10-15; ethylene dichloride 15-20; ethylene acetate 30-40; 95% ethylene alcohol 60-70; water 60-70.

Legal Information

Amberlyst is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acid-catalyzed conversion of furfuryl alcohol to ethyl levulinate in liquid ethanol.
Maldonado GMG, et al.
Energy & Environmental Science, 5(10), 8990-8997 (2012)
Biomass conversion to diesel via the etherification of furanyl alcohols catalyzed by Amberlyst-15.
Sacia, et al.
J. Catal., 313, 70-79 (2014)
A recyclable Amberlyst-15-catalyzed threecomponent reaction in water to synthesize
diarylmethyl sulfones
Kuchukulla RR, et al.
Green Chemistry, 21, 5808-5812 (2019)
Ion-exchange resins as catalysts in transesterification of triolein.
Paterson G, et al.
Catalysis Today, 212(10), 157-163 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service