Skip to Content
Merck
All Photos(3)

Key Documents

518018

Sigma-Aldrich

Lithium iodide

greener alternative

99.9% trace metals basis

Synonym(s):

Lithium(1+)iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiI
CAS Number:
Molecular Weight:
133.85
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

Quality Level

Assay

99.9% trace metals basis

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤1500.0 ppm Trace Metal Analysis

mp

446 °C (lit.)

density

3.49 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

[Li+].[I-]

InChI

1S/HI.Li/h1H;/q;+1/p-1

InChI key

HSZCZNFXUDYRKD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithium iodide is widely used as an electrolyte additivein dye-sensitized solar cells and Li-S batteries, as it enables long cyclelife. It is also used as a phosphor for neutron detection.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Lithium iodide(LiI)can be used as an electrolyte additive:

  • To prepare biodegradable polymer electrolytes.Rice starch complexed with LiI shows enhanced ionic conductivity as theaddition of LiI increases the number of mobile charge carriers.
  • For Li-S batteries. LiIforms a protective coating on the surface of both negative and positiveelectrodes and prevents the dissolution of polysulfides on the cathode sidewhich significantly enhances cell rate performance.
It can also be used to prepare Li-basedscintillators with enhanced thermal neutron detection efficiency.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feixiang Wu et al.
Advanced materials (Deerfield Beach, Fla.), 27(1), 101-108 (2014-11-05)
Lithium Iodide (LiI) is reported as a promising electrolyte additive for lithium-sulfur batteries. It induces formation of Li-ion-permeable protective coatings on both positive and negative electrodes, which prevent the dissolution of polysulfides on the cathode and reduction of polysulfides on
Seon-Joo Choi et al.
ACS applied materials & interfaces, 10(37), 31404-31412 (2018-08-28)
All-solid-state lithium batteries (ASSLBs) based on sulfide solid electrolytes (SEs) have received great attention because of the high ionic conductivity of the SEs, intrinsic thermal safety, and higher energy density achievable with a Li metal anode. However, studies on practical
Yu-il Kang et al.
ChemSusChem, 8(22), 3799-3804 (2015-10-17)
Dye-sensitized solar cells (DSCs) with long-term stability are produced using polymer-gel electrolytes (PGEs). In this study, we introduce the formation of PGEs using in situ gelation with poly(methyl methacrylate) (PMMA) particles and graphene fillers that are pre-deposited on the counter electrodes.
Jianjian Lin et al.
Scientific reports, 4, 5769-5769 (2014-08-30)
Three-dimensional (3D) hierarchical nanoscale architectures comprised of building blocks, with specifically engineered morphologies, are expected to play important roles in the fabrication of 'next generation' microelectronic and optoelectronic devices due to their high surface-to-volume ratio as well as opto-electronic properties.
Jung-Che Tsai et al.
Chemistry, an Asian journal, 10(9), 1932-1939 (2015-07-15)
Mesoporous cobalt sulfide nanotube arrays on FTO-coated glass were synthesized by combining three simple technologies: the selective etching of ZnO sacrificial templates, mesoporous Co3 O4 formation from cobalt-chelated chitosan, and ion-exchange reaction (IER). The mesoporous Co3 O4 nanotubes composed of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service