Skip to Content
Merck
All Photos(2)

Documents

427152

Sigma-Aldrich

Poly(vinylidene fluoride)

average Mw ~180,000 by GPC, average Mn ~71,000, beads or pellets

Synonym(s):

PVDF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CF2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

15 mmHg ( 32 °C)

form

beads or pellets

melt index

20-35 g/10 min (230°C/3.8kg)

mol wt

average Mn ~71,000
average Mw ~180,000 by GPC

impact strength

2-4 ft-lb/in. (Izod, ASTM D 256, notched)

dielectric constant

8.0-9.5, 100 Hz (ASTM D 150)

hardness

76-80 (Shore D, ASTM D 2240)

refractive index

n20/D 1.42

ASTM D 3835

6,000-7,500 poise(230 °C) (100 sec-1)(lit.)

transition temp

brittleness temperature -62 °C (ASTM D 746)
Tm 155-165 °C

solubility

some polar solvents such as organic esters and amines: soluble

density

1.78 g/mL at 25 °C

SMILES string

FC(F)=C

InChI

1S/C2H2F2/c1-2(3)4/h1H2

InChI key

BQCIDUSAKPWEOX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Coatings, film, filter cloth, instrument linings, filtration membranes, pump parts and linings and valves.

Features and Benefits

Crystallizes in at least 3 forms, depending on crystallization conditions. High mechanical strength,abrasion resistance and dielectric strength. Resists most chemicals, UV radiation, weathering and fungi.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Daisuke Matsutomo et al.
Journal of biochemistry, 153(2), 179-190 (2012-11-07)
Diacylglycerol kinase (DGK) participates in regulating the intracellular concentrations of two bioactive lipids, diacylglycerol and phosphatidic acid. DGKη1 is a type II isozyme that contains a pleckstrin homology (PH) domain and a pair of C1 domains at the N-terminus and
Eugene Cho et al.
Journal of nanoscience and nanotechnology, 13(1), 674-677 (2013-05-08)
Electrospun polyvinylidene fluoride (PVDF) nanofiber membranes have 3-dementional (3-D) open pore channel and hence have excellent application potential in Western blot. In this study we have modified electrospun PVDF nanofiber membrane by argon (Ar) plasma treatment to improve the surface
Noriko Kotobuki et al.
Journal of biomedical materials research. Part A, 101(2), 537-546 (2012-08-29)
For tissue engineering and regenerative medicine, stem cells should be effectively cultured in vitro. New thermoresponsive nanocomposite gels (MD-NC gels), consisting of inorganic clay (hectorite) and copolymers composed of hydrophobic 2-methoxyethyl acrylate (MEA) and hydrophilic N,N-dimethylacrylamide (DMAA) units, could be
S V Ivanov et al.
Klinichna khirurhiia, (7)(7), 37-41 (2012-10-05)
The results of experimental and clinical studies for the comparative evaluation of implants used in herniology of polytetrafluoroethylene (PTFE) and polyvinylidene fluoride (PVDF) are presented. Experimental investigations conducted on 200 mice, which in the anterior abdominal wall mesh prosthesis implanted.
Orang Seyeddain et al.
Journal of cataract and refractive surgery, 39(2), 234-241 (2012-12-19)
To report the 2-year postoperative safety and efficacy outcomes after monocular Kamra corneal inlay (ACI7000PDT) implantation in femtosecond laser-created corneal pockets of emmetropic presbyopic patients to improve near and intermediate vision. University Eye Clinic, Paracelsus Medical University, Salzburg, Austria. Prospective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service