Skip to Content
Merck
All Photos(1)

Key Documents

418951

Sigma-Aldrich

(DHQD)2Pyr

97%

Synonym(s):

Hydroquinidine-2,5-diphenyl-4,6-pyrimidinediyl diether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H60N6O4
CAS Number:
Molecular Weight:
881.11
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

optical activity

[α]20/D −390°, c = 1.2 in methanol

mp

247-250 °C (lit.)

functional group

phenyl

SMILES string

CC[C@H]1CN2CCC1CC2[C@@H](Oc3nc(nc(O[C@H](C4CC5CCN4C[C@@H]5CC)c6ccnc7ccc(OC)cc67)c3-c8ccccc8)-c9ccccc9)c%10ccnc%11ccc(OC)cc%10%11

InChI

1S/C56H60N6O4/c1-5-35-33-61-27-23-39(35)29-49(61)52(43-21-25-57-47-19-17-41(63-3)31-45(43)47)65-55-51(37-13-9-7-10-14-37)56(60-54(59-55)38-15-11-8-12-16-38)66-53(50-30-40-24-28-62(50)34-36(40)6-2)44-22-26-58-48-20-18-42(64-4)32-46(44)48/h7-22,25-26,31-32,35-36,39-40,49-50,52-53H,5-6,23-24,27-30,33-34H2,1-4H3/t35-,36-,39-,40-,49+,50+,52-,53-/m0/s1

InChI key

SWKRDCRSJPRVNF-DOGDSVMGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(DHQD)2Pyr may be used:
  • To catalyze the enantioselective allylic amination of alkylidene cyanoacetates with dialkyl azodicarboxylates.
  • To prepare 10(R),11-Dihydroxydihydroquinine and 10(S),11-dihydroxydihydroquinine via the Sharpless catalytic asymmetric dihydroxylation.
  • In the total synthesis of okadaic acid, a protein phosphatase inhibitor.

Legal Information

Sold under license from Rhodia Pharma Solutions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Total synthesis of the protein phosphatase inhibitor okadaic acid.
Ley SV, et al.
Journal of the Chemical Society. Perkin Transactions 1, 23, 3907-3912 (1998)
Enantioselective organocatalytic allylic amination.
Journal of the American Chemical Society, 127(33), 11614-11615 (2005)
Stereocontrolled conversion of quinine into 10 (R), 11-dihydroxydihydroquinine via the sharpless osmylation process.
Zheng T, et al.
The Journal of Organic Chemistry, 66(4), 1509-1511 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service