Skip to Content
Merck
All Photos(2)

Documents

209104

Sigma-Aldrich

Potassium osmate(VI) dihydrate

Synonym(s):

Dipotassium tetrahydroxodioxoosmate, Potassium dioxidodioxoosmium dihydrate, Potassium osmium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2OsO4·2H2O
CAS Number:
Molecular Weight:
368.45
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

form

powder

Quality Level

reaction suitability

reagent type: oxidant

SMILES string

O.O.[K+].[K+].[O-][Os]([O-])(=O)=O

InChI

1S/2K.2H2O.4O.Os/h;;2*1H2;;;;;/q2*+1;;;;;2*-1;

InChI key

DGODWNOPHMXOTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Potassium osmate(VI) dihydrate is a hydroscopic transition metal compound soluble in water and methanol, widely used as a reagent/catalyst to synthesize Os compounds.

Application

Potassium osmate(VI) dihydrate can be used:
  • As a reagent for the stereoselective synthesis of 2,3,4-unprotected β-N-Glycopyranosides via palladium-catalyzed Tsuji-Trost amination.
  • As a catalyst for asymmetric synthesis of carboranylated diols bearing two adjacent stereocenters.
  • To prepare Osmium complexes for voltammetric analysis of polysaccharides.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy J Donohoe et al.
Journal of the American Chemical Society, 128(8), 2514-2515 (2006-02-24)
The use of N-sulfonyloxy carbamates as reoxidants for the tethered aminohydroxylation (TA) reaction is reported. These new conditions obviate the requirement for lithium hydroxide and tBuOCl in the oxidation mixture. In addition to providing aminohydroxylation products in good yields, the
Tetrahedron Letters, 47, 3797-3797 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service