Skip to Content
Merck
All Photos(2)

Key Documents

400203

Sigma-Aldrich

Lithium ethoxide

greener alternative

95%

Synonym(s):

LiOEt, Lithium ethanolate, Lithium ethylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OLi
CAS Number:
Molecular Weight:
52.00
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

powder and chunks

reaction suitability

core: lithium

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

[Li+].CC[O-]

InChI

1S/C2H5O.Li/c1-2-3;/h2H2,1H3;/q-1;+1

InChI key

AZVCGYPLLBEUNV-UHFFFAOYSA-N

General description

Lithium ethoxide is an organometallic compound widely used as a precursor to fabricate lithium-ion batteries.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Lithium ethoxide can be used as a precursor:
  • To fabricate chemically inert, Li-ion conducting, and electronically insulating coating on the cathode materials. This coating helps to avoid unwanted reactions between the solid-state electrolyte and the cathode in all-solid-state batteries.
  • To produce an amorphous Li-Zr-O coating on the surface of high-Ni cathodes, to improve the surface stability and cycling performance of lithium-ion batteries.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Self-heat. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amorphous Li-Zr-O layer coating on the surface of high-Ni cathode materials for lithium ion batteries
Young Jin Lim, et al.
Electrochimica Acta, 282, 311-316 (2018)
A facile, dry-processed lithium borate-based cathode coating for improved all-solid-state battery performance
Erik A Wu, et al.
Journal of the Electrochemical Society, 167, 130516-130516 (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service