Skip to Content
Merck
All Photos(1)

Key Documents

398195

Sigma-Aldrich

Lithium tert-butoxide solution

1.0 M in THF

Synonym(s):

tert-Butoxylithium, Lithium 2-methylpropan-2-olate, Lithium t-butoxide, Lithium tert-butylate, Lithium salt of tert-Butyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COLi
CAS Number:
Molecular Weight:
80.05
Beilstein:
3620018
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

concentration

1.0 M in THF

bp

67 °C

density

0.888 g/mL at 25 °C

SMILES string

[Li+].CC(C)(C)[O-]

InChI

1S/C4H9O.Li/c1-4(2,3)5;/h1-3H3;/q-1;+1

InChI key

LZWQNOHZMQIFBX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Lithium tert-butoxide solution is generally used as a strong base in organic synthesis.
It can be used:
  • For the synthesis of lithium modified silica nano-particles for conductive gel electrolytes.
  • As a catalyst for ring-opening polymerization of lactides.
  • As a lithium precursor for the synthesis of LiV3O8 nanoparticles by flame spray pyrolysis.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Self-heat. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient and controlled polymerization of lactide under mild conditions with a sodium-based catalyst.
Chen HY, et al.
Green Chemistry, 9(10), 1038-1040 (2007)
Solid-phase synthesis of quinoxaline, thiazine, and oxazine analogs through a benzyne intermediate.
Dixon S, et al.
Tetrahedron Letters, 46(43), 7443-7446 (2005)
Electrochemistry of LiV3O8 nanoparticles made by flame spray pyrolysis.
Patey TJ, et al.
Electrochemical and Solid-State Letters, 11(4), A46-A50 (2008)
Gel electrolytes based on lithium modified silica nano-particles.
Sun J, et al.
Electrochimica Acta, 52(24), 7083-7090 (2007)
Teng-Hao Chen et al.
Nature communications, 5, 5131-5131 (2014-10-14)
Metal-organic and covalent organic frameworks are porous materials characterized by outstanding thermal stability, high porosities and modular synthesis. Their repeating structures offer a great degree of control over pore sizes, dimensions and surface properties. Similarly precise engineering at the nanoscale

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service