Skip to Content
Merck
All Photos(1)

Documents

385565

Sigma-Aldrich

Methyl 8-chloro-8-oxooctanoate

96%

Synonym(s):

Methyl suberyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCO(CH2)6CO2CH3
CAS Number:
Molecular Weight:
206.67
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.45 (lit.)

density

1.456 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCCCCCC(Cl)=O

InChI

1S/C9H15ClO3/c1-13-9(12)7-5-3-2-4-6-8(10)11/h2-7H2,1H3

InChI key

RKUPOLBFJIEWBZ-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A cyclodextrin-capped histone deacetylase inhibitor.
Amin et al.
Biochemical Medicine, 23(11), 3346-3348 (2013)
Synthesis and properties of PI polyamide?SAHA conjugate.
Ohtsuki et al.
Tetrahedron Letters, 50(52), 7288-7292 (2009)
Synthesis of 1-Aminocyclopropyl-Pentanoic and-Heptanoic Acid Derivatives.
Gensini et al.
Letters in Organic Chemistry, 5(5), 328-331 (2008)
Laura Forster et al.
Analytical and bioanalytical chemistry, 394(6), 1679-1685 (2009-06-02)
A fluorescent assay for the evaluation of inhibitors of fatty acid amide hydrolase (FAAH) is described. Microsomes from rat brain served as enzyme source. N-(2-Hydroxyethyl)-4-pyren-1-ylbutanamide was designed and synthesized as novel fluorogenic substrate. For substrate solubilization, Triton X-100 was employed.
John Spencer et al.
ACS medicinal chemistry letters, 2(5), 358-362 (2011-05-17)
N(1)-Hydroxy-N(8)-ferrocenyloctanediamide, JAHA (7), an organometallic analogue of SAHA containing a ferrocenyl group as a phenyl bioisostere, displays nanomolar inhibition of class I HDACs, excellent selectivity over class IIa HDACs, and anticancer action in intact cells (IC(50) = 2.4 μM, MCF7

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service