Skip to Content
Merck
All Photos(1)

Key Documents

365963

Sigma-Aldrich

Di-tert-butyl N,N-diethylphosphoramidite

technical grade, 93%

Synonym(s):

Bis(1,1-dimethylethyl) N,N-diethylphosphoramidite, Di-tert-butyl diethylphosphoramidite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NP[OC(CH3)3]2
CAS Number:
Molecular Weight:
249.33
Beilstein:
4175262
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

93%

form

liquid

refractive index

n20/D 1.434 (lit.)

bp

39-41 °C/0.3 mmHg (lit.)

density

0.896 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCN(CC)P(OC(C)(C)C)OC(C)(C)C

InChI

1S/C12H28NO2P/c1-9-13(10-2)16(14-11(3,4)5)15-12(6,7)8/h9-10H2,1-8H3

InChI key

KUKSUQKELVOKBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calcium phosphate nanoparticles: colloidally stabilized and made fluorescent by a phosphate-functionalized porphyrin.
Ganesan K, et al.
Journal of Materials Chemistry, 18(31), 3655-3661 (2008)
J W Perich et al.
Bioorganic & medicinal chemistry, 4(2), 143-150 (1996-02-01)
The seven phosphopeptide derivatives based on the native -NEYTA- sequence of the pp60src protein kinase family, Asn-Glu-Tyr(P)-Ser-Ala, Ala-Glu-Tyr(P)-Ser-Ala, Ala-Ser-Tyr(P)-Ser-Ala, Ala-Ser(P)-Tyr-Ser-Ala, Ala-Thr-Tyr(P)-Ser-Ala, Ala-Thr(P)-Tyr-Ser-Ala and Ala-Ser(P)-Tyr(P)-Ser-Ala, were prepared in good yield using the "global' "phosphite-triester' phosphorylation method. The peptide resins were assembled
H Mostafavi et al.
Biomedical peptides, proteins & nucleic acids : structure, synthesis & biological activity, 1(5), 255-260 (1995-01-01)
Based on the global phosphorylation approach, a selective synthesis of (Ser10-phosphatidyl)-urodilatin (phosphourodilatin), which contains 32 amino acid residues and a disulfide loop is described. The peptide was assembled stepwise on a polyethyleneglycol-polystyrene support using Fmoc-chemistry. The phosphorylation was performed on-resin
D Dunn et al.
The Journal of biological chemistry, 271(1), 168-173 (1996-01-05)
Yersinia protein-tyrosine phosphatase substrates have been synthesized employing an expedient methodology that incorporates phosphorylated non-amino acid residues into an active site-directed peptide. While the peptidic portion of these compounds serves an enzyme targeting role, the nonpeptidic component provides a critical

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service