
Di-tert-butyl N,N-diethylphosphoramidite

Linear Formula:
(C2H5)2NP[OC(CH3)3]2
CAS Number:
Molecular Weight:
249.33
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing