Merck
  • Ultrasmall-Superbright Neodymium-Upconversion Nanoparticles via Energy Migration Manipulation and Lattice Modification: 808 nm-Activated Drug Release.
Ultrasmall-Superbright Neodymium-Upconversion Nanoparticles via Energy Migration Manipulation and Lattice Modification: 808 nm-Activated Drug Release.
ACS nano (2017-02-22)
Yan Zhang, Zhongzheng Yu, Jingqiu Li, Yanxiao Ao, Jingwen Xue, Zhiping Zeng, Xiangliang Yang, Timothy Thatt Yang Tan
ABSTRACT
Nd
Sigma-Aldrich
Neodymium(III) acetate hydrate, 99.9%
Sigma-Aldrich
Thulium(III) acetate hydrate, 99.9% trace metals basis
Sigma-Aldrich
Ytterbium(III) acetate hydrate, 99.95% trace metals basis
Sigma-Aldrich
Gadolinium(III) acetate hydrate, 99.9% trace metals basis
Sigma-Aldrich
Ruthenium(III) chloride trihydrate, technical
Sigma-Aldrich
Oleic acid, technical grade, 90%
Sigma-Aldrich
1-Octadecene, technical grade, 90%