357626
Nickel
wire, diam. 0.5 mm, ≥99.9% trace metals basis
Synonym(s):
Ni
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥99.9% trace metals basis
form
wire
reaction suitability
core: nickel
reagent type: catalyst
resistivity
6.97 μΩ-cm, 20°C
diam.
0.5 mm
bp
2732 °C (lit.)
mp
1453 °C (lit.)
density
8.9 g/mL at 25 °C (lit.)
SMILES string
[Ni]
InChI
1S/Ni
InChI key
PXHVJJICTQNCMI-UHFFFAOYSA-N
Related Categories
Quantity
- 5 m (approximately 8.5 g)
- 25 m (approximately 42.5 g)
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Skin Sens. 1 - STOT RE 1
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Ornella Valenti et al.
Cerebral cortex (New York, N.Y. : 1991), 19(3), 658-674 (2008-07-18)
The prefrontal cortex receives multiple inputs from the hippocampal complex, which are thought to drive memory-guided behavior. Moreover, dysfunctions of both regions have been repeatedly associated with several psychiatric disorders. Therefore, understanding the interconnections and modulatory interactions between these regions
A J Feilzer et al.
Nederlands tijdschrift voor tandheelkunde, 120(6), 335-341 (2013-07-19)
Orally applied metal alloys can cause undesirable physical effects. A distinction needs to be made in this respect between local and systemic reactions and toxic and immunological reactions. A case is presented which illustrates this problem. In this case, the
Fariborz Amini et al.
International journal of orthodontics (Milwaukee, Wis.), 24(1), 43-44 (2013-06-05)
In this report, we present an uncomplicated approach to overcome lingual rolling of mandibular molars during orthodontic treatment, by using rectangular NiTi arch wire in twist fashion through the molar tube. The sequential steps of the twist manoeuvre on a
Palatally anchored maxillary molar mesialization using the mesialslider.
Benedict Wilmes et al.
Journal of clinical orthodontics : JCO, 47(3), 172-179 (2013-05-11)
Naeemullah et al.
Journal of AOAC International, 96(2), 447-452 (2013-06-19)
A simple method for the preconcentration of cadmium (Cd) and nickel (Ni) in drinking and wastewater samples was developed. Cloud point extraction has been used for the preconcentration of both metals, after formation of complexes with 8-hydroxyquinoline (8-HQ) and extraction
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service