Skip to Content
Merck
All Photos(1)

Key Documents

267007

Sigma-Aldrich

Nickel

foil, thickness 0.25 mm, 99.995% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ni
CAS Number:
Molecular Weight:
58.69
EC Number:
MDL number:
UNSPSC Code:
12141729
PubChem Substance ID:
NACRES:
NA.23

Assay

99.995% trace metals basis

form

foil

resistivity

6.97 μΩ-cm, 20°C

thickness

0.25 mm

bp

2732 °C (lit.)

mp

1453 °C (lit.)

density

8.9 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Ni]

InChI

1S/Ni

InChI key

PXHVJJICTQNCMI-UHFFFAOYSA-N

General description

Nickel foil has been used as a substrate for the growth of several nanomaterials. A superhydrophobic surface of nanowire structure was grown on a nickel foil by solution immersion method. Co2O4 nanorods may be grown on nickel foil by hydrothermal reaction. A study reports the growth of carbon nanofibers on nickel, as a catalyst by chemical vapor deposition (CVD). Nickel foils may also act as base for (Pb0.92La0.08)(Zr0.52Ti0.48)O3 (PLZT) films.

Quantity

1.4 g = 25 × 25 mm, 5.6 g = 50 × 50 mm

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1 - STOT RE 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wettability of carbon nanofiber layers on nickel foils
Pacheco Benito S, et al.
Journal of Colloid and Interface Science, 364(2), 530-538 (2011)
Facile fabrication of superhydrophobic surface with nanowire structures on nickel foil,
Zhang X, et al.
Applied Surface Science, 287, 299-303 (2013)
Effects of barrier layers on the electrical properties of (Pb,La)(Zr,Ti)O3 films on nickel foils
Kim DJ, et al.
Materials Science and Engineering, B, 141(1-2), 87-90 (2007)
Hydrothermal synthesis of Co3O4 nanorods on nickel foil
Dong C, et al.
Materials Letters, 123, 187-190 (2014)
Ornella Valenti et al.
Cerebral cortex (New York, N.Y. : 1991), 19(3), 658-674 (2008-07-18)
The prefrontal cortex receives multiple inputs from the hippocampal complex, which are thought to drive memory-guided behavior. Moreover, dysfunctions of both regions have been repeatedly associated with several psychiatric disorders. Therefore, understanding the interconnections and modulatory interactions between these regions

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service