Skip to Content
Merck
All Photos(1)

Key Documents

343633

Sigma-Aldrich

4,4,4-Trifluoro-1-(2-naphthyl)-1,3-butanedione

99%

Synonym(s):

4,4,4-Trifluoro-3-oxo-2′-butyronaphthone, 1-(2-Naphthoyl)-3,3,3-trifluoroacetone, 2-(4,4,4-Trifluoroacetoacetyl)naphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7COCH2COCF3
CAS Number:
Molecular Weight:
266.22
Beilstein:
1981085
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

form

powder

mp

70-72 °C (lit.)

SMILES string

FC(F)(F)C(=O)CC(=O)c1ccc2ccccc2c1

InChI

1S/C14H9F3O2/c15-14(16,17)13(19)8-12(18)11-6-5-9-3-1-2-4-10(9)7-11/h1-7H,8H2

InChI key

WVVLURYIQCXPIV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daewon Han et al.
Nanomaterials (Basel, Switzerland), 9(1) (2019-01-10)
Red emitting europium (III) complexes Eu(TFAAN)₃(P(Oct)₃)₃ (TFAAN = 2-(4,4,4-Trifluoroacetoacetyl)naphthalene, P(Oct)₃ = trioctylphosphine) chelated on carboxymethyl dextran coated superparamagnetic iron oxide nanoparticles (CMD-SPIONs) was synthesized and the step wise synthetic process was reported. All the excitation spectra of distinctive photoluminesces were
Eliot Morrison et al.
Communications biology, 3(1), 368-368 (2020-07-12)
Palmitoylation is the reversible addition of palmitate to cysteine via a thioester linkage. The reversible nature of this modification makes it a prime candidate as a mechanism for regulating signal transduction in T-cell receptor signaling. Following stimulation of the T-cell
Longjun Wang et al.
Steroids, 89, 41-46 (2014-08-06)
An ultrasensitive and selective method is presented for the determination of diethylstilbestrol (DES) using time-resolved fluorescence immunoassay (TRFIA) based on double-codified gold nanoparticles (DC-AuNPs). In this system, the DC-AuNPs, that are gold nanoparticles (AuNPs) modified with anti-DES antibody and SH-dsDNA-biotin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service