Skip to Content
Merck
All Photos(1)

Key Documents

325384

Sigma-Aldrich

N-Methylsuccinimide

99%

Synonym(s):

1-Methyl-2,5-pyrrolidinedione, N-Methyl-2,5-pyrrolidinedione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7NO2
CAS Number:
Molecular Weight:
113.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

bp

234-235 °C (lit.)

mp

61-70 °C (lit.)

SMILES string

CN1C(=O)CCC1=O

InChI

1S/C5H7NO2/c1-6-4(7)2-3-5(6)8/h2-3H2,1H3

InChI key

KYEACNNYFNZCST-UHFFFAOYSA-N

General description

N-Methylsuccinimide is a metabolite of N-methyl-2-pyrrolidone (NMP) and its presence in in plasma and urine can be used as a biomarker of exposure to NMP. Hydrogen bonded complexes between N-methylsuccinimide and phenols (pKa = 10.2 → 6) are investigated by infrared spectrometry.

Application

N-Methylsuccinimide was employed as model compound to investigate the mechanism of enolization step by density-functional theory (DFT) calculations.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-Methylsuccinimide in plasma and urine as a biomarker of exposure to N-methyl-2-pyrrolidone.
Jonsson BAG and ?kesson B.
International Archives of Occupational and Environmental Health, 74(4), 289-294 (2004)
nfrared Study of the Hydrogen Bond Complexes Between N-Methylsuccinimide and Phenols.
Bruyneel K, et al.
Spectroscopy Letters, 29(4), 739-747 (1996)
Ohgi Takahashi et al.
Chemistry & biodiversity, 7(6), 1349-1356 (2010-06-22)
Racemization of aspartic acid residues in peptides and proteins is assumed to proceed via succinimide intermediates. An enolization of the succinimide intermediate is required for the racemization to occur. In this study, we modeled the enolization step by density-functional theory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service