Skip to Content
Merck
All Photos(1)

Key Documents

311294

Sigma-Aldrich

1,4,7-Trimethyl-1,4,7-triazacyclononane

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H21N3
CAS Number:
Molecular Weight:
171.28
Beilstein:
4174812
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.473 (lit.)

density

0.884 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CN1CCN(C)CCN(C)CC1

InChI

1S/C9H21N3/c1-10-4-6-11(2)8-9-12(3)7-5-10/h4-9H2,1-3H3

InChI key

WLDGDTPNAKWAIR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Useful ligand for preparing transition metal complexes.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

152.6 °F - closed cup

Flash Point(C)

67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inorganic Chemistry, 33, 3990-3990 (1994)
Journal of the Chemical Society. Chemical Communications, 1780-1780 (1992)
Journal of the American Chemical Society, 114, 10676-10676 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service