Skip to Content
Merck
All Photos(2)

Key Documents

259160

Sigma-Aldrich

1,4,8,11-Tetraazacyclotetradecane

98% (GC)

Synonym(s):

Cyclam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H24N4
CAS Number:
Molecular Weight:
200.32
Beilstein:
111811
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98% (GC)

mp

184-186 °C (lit.)

SMILES string

C1CNCCNCCCNCCNC1

InChI

1S/C10H24N4/c1-3-11-7-9-13-5-2-6-14-10-8-12-4-1/h11-14H,1-10H2

InChI key

MDAXKAUIABOHTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4,8,11-Tetraazacyclotetradecane is an azamacrocycle that forms more stable metal complexes as compared to open chain ligands.

Application

1,4,8,11-Tetraazacyclotetradecane (cyclam) can be used as a ligand in the synthesis of:
  • Cyanide-bridged FeIII-CuII complexes.
  • Zinc fenamate complexes [Zn(cyclam)(fen)2] possessing antimicrobial properties, fen = fenamic acid.
  • Zn(II) complexes with flufenamic acid (flu), [Zn(cyclam)(flu)2].
  • Polyoxometalate-metal organic extended framework, for example [{Cu(cyclam)}3(W7O24)].15.5.H2O.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclam (1,4,8,11-tetraazacyclotetradecane) with one methylphosphonate pendant arm: a new ligand for selective copper (II) binding.
F?zerova S, et al.
Dalton Transactions, 2908-2915 (2005)
Abir El Majzoub et al.
Inorganic chemistry, 50(9), 4029-4038 (2011-04-12)
The coordination properties and the photophysical response of a new cyclam fluorescent probe for Zn(II), [L1H: 1-(benzimidazol-2-ylmethyl]-1,4, 8,11-tetraazacyclotetradecane] toward Cu(II), Zn(II), and Cd(II) are reported. The stability constants of the corresponding complexes were determined by means of potentiometric measurements in
Mingfeng Yu et al.
Inorganic chemistry, 50(24), 12823-12835 (2011-11-25)
We describe the synthesis of cyclam metal complexes derivatized with amino acids or a tripeptide using a copper(I)-catalyzed Huisgen "click" reaction. The linker triazole formed during the synthesis plays an active coordinating role in the complexes. The reaction conditions do
Jing Li et al.
Biomacromolecules, 13(10), 3220-3227 (2012-09-26)
A series of reducible polycationic copper chelators (RPCs) based on 1,4,8,11-tetraazacyclotetradecane (cyclam) were synthesized by Michael addition. Molecular weight of the polycations was controlled by reaction stoichiometry and reaction conditions, resulting in polymers with molecular weights ranging from 4400 to
Apurba Kalita et al.
Inorganic chemistry, 50(23), 11868-11876 (2011-11-02)
The nitric oxide reactivity of two copper(II) complexes, 1 and 2 with ligands L(1) and L(2), respectively, [L(1) = 5,5,7,12,12,14-hexamethyl-1,4,8,11-tetraazacyclotetradecane, L(2) = 5,5,7-trimethyl-[1,4]-diazepane] have been studied. The copper(II) center in complex 1 was found to be unreactive toward nitric oxide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service