Skip to Content
Merck
All Photos(1)

Key Documents

309230

Sigma-Aldrich

Thallium(I) nitrate

99.9% trace metals basis

Synonym(s):

Thallium mononitrate, Thallous nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
TlNO3
CAS Number:
Molecular Weight:
266.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid
solid

Quality Level

Assay

99.9% trace metals basis

form

solid
solid

impurities

≤1500 mg/kg Trace metallic impurities analysis (ICP)

mp

206 °C (lit.)

SMILES string

[Tl+].[O-][N+]([O-])=O

InChI

1S/NO3.Tl/c2-1(3)4;/q-1;+1

InChI key

FYWSTUCDSVYLPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Thallium(I) nitrate is a white crystalline solidwith strong oxidizing properties. It is widely applied in the field of organicsynthesis, analytical chemistry, and electronics.

Application

Thallium(I) nitrate can be used:
  • As an additive to synthesize thallium-containing cadmium selenide thin films by electrochemical deposition. The addition of TlNO3 decreases corrosion and helps to form uniform and stable thin films.
  • As a dopant to prepare bismuth vanadate nanoparticles. TlNO3 reduces charge carrier recombination and enhances the photocatalytic activity.
  • To prepare precursor solution for the detection of halogens by molecular absorption spectroscopy.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - Ox. Sol. 3 - STOT RE 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of bromine and fluorine via sequential vaporization of TlBr and CaF molecules from the same aliquot by high-resolution continuum source graphite furnace molecular absorption spectrometry
Amauri P.A. Rosa and Tatiane de A. Maranh?o},
Spectrochimica Acta. Part B: Atomic Spectroscopy, 197, 106525-106525 (2022)
Enhanced visible light-photocatalysis by hydrothermally synthesized thallium-doped bismuth vanadate nanoparticles
C.Karunakaran and S.Kalaivani
Materials Science in Semiconductor Processing, 27, 352-361 (2014)
Vânia M T Carneiro et al.
The Journal of organic chemistry, 75(9), 2877-2882 (2010-04-10)
A diastereoselective route to (+)-bakkenolide A is presented from the readily available optically active Wieland-Miescher ketone. This novel synthesis of this sesquiterpene lactone features the following as key stereoselective transformations: (i) the ring contraction reaction of a octalone mediated by
Cecilia Eliana Hanzel et al.
Toxicology and applied pharmacology, 236(1), 59-70 (2009-04-18)
Thallium (Tl) is a highly toxic metal though yet its mechanisms are poorly understood. Previously, we demonstrated that rat pheochromocytoma (PC12) cells exposure to thallous (Tl(I)) or thallic (Tl(III)) cations leads to mitochondrial damage and reduced cell viability. In the
P Thanigaimalai et al.
Bioorganic & medicinal chemistry, 18(12), 4441-4445 (2010-05-18)
The oxidative cyclization of 2'-hydroxy-6'-cyclohexylmethoxychalcones 5 using thallium (III) nitrate (TTN) in alcoholic solvents produced isoflavones 2 and (or) aurones 3 depending on the electronic nature of p-substituents on ring B. Chalcones with strong electron donating substituents (OH, OCH(3)) were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service